missing translation for 'onlineSavingsMsg'
Learn More

Iodine 1 Normal, Safety Coated Glass, DEA List I, Reagents

Catalog No. 361059175
Change view
Click to view available options
Quantity:
4L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361059175 4L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
DEA LIST I
Catalog No. 361059175 Supplier Labchem Inc Supplier No. CI2025004C1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

DEA LIST I
  • Iodine, 1.0 N
  • DEA List I
  • Appearance: Dark Brown Liquid
  • Size: 4 L
  • Specific Gravity: 1.3
  • Brand: Reagents
  • NIST Traceable
  • Prepared with ACS Grade Components and ASTM Type II Water
  • Assay (vs. Sodium Thiosulfate/Starch) Spec: 0.995 to 1.005 N

Specifications

Physical Form Liquid or Solid
Quantity 4L
Molecular Weight (g/mol) Blend
Percent Purity 1.0 N
Grade Laboratory
Identification 1 L
Hazard Category Blend
Hazard Statement 1.0 N
Precautionary Statement Liquid or Solid