missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodide Standard For Ion Chromatography, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$326.22 - $689.66
Chemical Identifiers
| CAS | 7681-82-5 |
|---|---|
| Molecular Formula | INa |
| Molecular Weight (g/mol) | 149.894 |
| InChI Key | FVAUCKIRQBBSSJ-UHFFFAOYSA-M |
| PubChem CID | 5238 |
| ChEBI | CHEBI:33167 |
| IUPAC Name | sodium;iodide |
| SMILES | [Na+].[I-] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| I- | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7681-82-5 | |
| 149.894 | |
| 5238 | |
| sodium;iodide |
| INa | |
| FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
| CHEBI:33167 | |
| [Na+].[I-] |
Specifications
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| Sodium iodide,distilled water,conductivity or of similar purity | |
| Ion Chromatography | |
| Standard Solution | |
| FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
| [Na+].[I-] | |
| sodium;iodide | |
| 125 mL | |
| CHEBI:33167 | |
| Iodide Standard, For Ion Chromatography |
Safety and Handling
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only