missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodide Standard For Ion Chromatography, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC ASI92X
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| I- | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Iodide Standard | |
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| Sodium iodide,distilled water,conductivity or of similar purity | |
| Ion Chromatography | |
| Standard Solution | |
| FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
| [Na+].[I-] | |
| sodium;iodide | |
| 500 mL | |
| CHEBI:33167 |
| For Ion Chromatography | |
| 7681-82-5 , 7732-18-5 | |
| 99.9,0.1 | |
| INa | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Certified Reference Material | |
| Sodium Iodide | |
| Water | |
| 149.894 | |
| 5238 | |
| 1000mg/L |
Chemical Identifiers
| 7681-82-5 | |
| 149.894 | |
| 5238 | |
| sodium;iodide |
| INa | |
| FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
| CHEBI:33167 | |
| [Na+].[I-] |
Safety and Handling
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only