missing translation for 'onlineSavingsMsg'
Learn More

Iodide Iodate concentrate, For 1L standard solution, 0.05 M I2 (0.1N), Honeywell Fluka™

Catalog No. p-7122098
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Catalog No. Quantity
6002663 1 Pc.
60046685 6 x 1 Ea.
2 options

Catalog No. 6002663

Supplier: Honeywell Chemicals 380611EA

Only null left

for 1L standard solution, 0.05 M I2 (0.1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7681-11-0 , 7553-56-2 , 7758-05-6 , 7732-18-5
Molecular Formula IK
MDL Number MFCD00011355
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES [K+].[I-]

Specifications

Quantity 1 Pc.
UN Number NONH for all modes of transport
Beilstein 3587194
IUPAC Name potassium iodide
Molecular Weight (g/mol) 166.00
Packaging Ampoule
Chemical Name or Material Iodide Iodate concentrate
Hazard Statement P305 + P351 + P338