Learn More
Invitrogen™ Zero Blunt™ TOPO™ PCR Cloning Kit, with One Shot™ MAX Efficiency DH5α-T1R E. coli
Offers the fastest and easiest method for high-efficiency (cloning of blunt-end PCR products amplified with proofreading thermostable polymerases
Supplier: Invitrogen™ K282020
Description
Kit includes linearized and topoisomerase I-activated pCR™-Blunt II-TOPO Vector for 5-minute benchtop ligations without ligase.
Selected Zero Blunt TOPO PCR Cloning Kits are available in combo kits combined with a PureLink™ Quick Plasmid Miniprep Kit (50 preps) for fast plasmid purification of your TOPO-cloned inserts for downstream analysis.
- ccdB gene for positive selection
- EcoR I sites flanking the PCR product insertion site for easy excision of inserts
- Kanamycin and Zeocin™ resistance genes for your choice of selection in E. coli
- SP6 promoter/primer site for in vitro RNA transcription and sequencing
- M13 forward and reverse primer sites for sequencing or PCR screening
Applications:
Cloning, PCR Cloning
Specifications
| DH5α | |
| Box 1: • Linearized and topoisomerase I-activated pCR™Blunt II-TOPO™ vector • Salt solution • dNTPs • Control template • M13 forward and reverse primers • Sterile water Store at -5 to -30°C. All reagents are stable for 6 months when properly stored. Box 2: • One Shot™ Chemically Competent or Electrocomp™ E. coli • S.O.C. medium • Supercoiled pUC19 control plasmid Store at -68 to -85°C. |
|
| One Shot | |
| SP6 | |
| Blunt DNA Cloning Vectors |
| Blunt TOPO™ | |
| Kit | |
| PCR Cloning Kit | |
| 25 Reactions |
For Research Use Only. Not for use in diagnostic procedures.