missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ X-Gal DFS Item

Histochemical substrate for β-galactosidase

$125.00 - $596.00

Specifications

Storage Requirements Store at -5° to -30°C
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
15520034 Life Technologies
LS15520034
100mg Each for $125.00
15520018 Life Technologies
LS15520018
1g Each for $596.00
Specifications

Description

This substrate yields a blue precipitate upon hydrolysis, making it suitable for use in immunoblotting or immunocytochemical assays. Bacteria containing active β-galactosidase produce blue colonies when grown on plates containing X-Gal. X-Gal is supplied as white crystals and is soluble in dimethylformamide.

  • Substrate Type: Beta-Gal Substrate
  • Detection Method: Colorimetric
  • Cell Permeability: Cell-Permeant
  • Substrate Properties: Chemical Substrate

Order Info

Approved for shipment at Room Temperature or on Wet Ice.

Description & Specifications

Specifications

Store at -5° to -30°C