missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ TAE Buffer, 10X

Compatible with agarose gels
$101.00 - $267.00
Specifications
| Buffer | Running Buffer |
|---|---|
| Concentration | 10 X |
| Content And Storage | Store at room temperature. |
| Format | Bottle |
| Gel Compatibility | Agarose Gels |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
15558042
![]()
|
Invitrogen™
15558042 |
1 L |
Each for $101.00
|
|
|||||
15558026
|
Invitrogen™
15558026 |
4 L |
Each for $267.00
|
|
|||||
Description
- UltraPure 10X TAE Buffer is sterile-filtered solution of 400 mM Tris-acetate and 10 mM EDTA
- TAE Buffer is most commonly used buffer for agarose DNA electrophoresis
- Supplied in 1 L plastic bottles/4 L/10 L stackable Cubitainer™ Box
- 1X TAE Buffer solution contains 40 mM Tris-acetate and 1mM EDTA at pH 8.3
- Performance and Quality Testing: No DNase activity detected. Sustainable packaging
Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
| Running Buffer | |
| Store at room temperature. | |
| Agarose Gels | |
| UltraPure | |
| Room Temperature |
| 10 X | |
| Bottle | |
| Nucleic Acid Gel Electrophoresis, Blotting | |
| TAE Buffer |
For Research Use Only. Not for use in diagnostic procedures.