missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ TAE Buffer, 10X
GREENER_CHOICE

Catalog No. 15558042
Encompass
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15558042 1 L
15558026 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15558042 Supplier Invitrogen™ Supplier No. 15558042

Compatible with agarose gels

  • UltraPure 10X TAE Buffer is sterile-filtered solution of 400 mM Tris-acetate and 10 mM EDTA
  • TAE Buffer is most commonly used buffer for agarose DNA electrophoresis
  • Supplied in 1 L plastic bottles/4 L/10 L stackable Cubitainer™ Box
  • 1X TAE Buffer solution contains 40 mM Tris-acetate and 1mM EDTA at pH 8.3
  • Performance and Quality Testing: No DNase activity detected. Sustainable packaging

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Buffer Running Buffer
Concentration 10 X
Content And Storage Store at room temperature.
Format Bottle
Gel Compatibility Agarose Gels
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Product Line UltraPure
Product Type TAE Buffer
Quantity 1 L
Shipping Condition Room Temperature
How can generation of replication competent adenoviruses be avoided when using your pSilencer adeno 1.0-CMV System?

Although there is only a very small chance of creating replication competent virus, steps should still be taken to avoid added risk. The virus is expanded in two rounds of amplification, and all secondary expansions should be performed from an initial expansion stock. Secondary amplifications in HEK293 cells should not be performed from the stock of another secondary expansion, as this could increase the chance of making replication competent virus. See the product manual for more detail and guidelines for screening.

For Research Use Only. Not for use in diagnostic procedures.