missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v)

UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v) is used in the purification of nucleic acids. This reagent consists of highly pure chloroform, isoamyl alcohol, and UltraPure™ Phenol saturated with Tris-HCl.

$274.00 - $743.00

Specifications

Chemical Name or Material Phenols
pH 8.05
Product Line UltraPure
Purity Molecular Biology Grade
Shipping Condition Room Temperature
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
15593031
Encompass
View Documents Promotion Details
Invitrogen™
15593031
100 mL
Each for $274.00
Only null left
 
15593049
Encompass
View Documents Promotion Details
Invitrogen™
15593049
400 mL
Each for $743.00
Only null left
 
Description

Description

  • Reagent consists of highly pure chloroform, isoamyl alcohol, and UltraPure™ Phenol saturated with Tris-HCl
  • When mixtures are extracted with Phenol:Chloroform:Isoamyl Alcohol, proteins are denatured and collected in the organic phase or at the interphase, while nucleic acids remain in the aqueous phase
  • UltraPure™ Phenol:Chloroform:lsoamyl Alcohol contains no preservatives
  • It is packaged under an inert gas in shatter-resistant, plastic-coated amber bottles
  • The appearance of the solution is evaluated at room temperature
  • No RNase or DNase activity is detected
  • Store in the refrigerator at 2°C to 8°C

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Phenols
UltraPure
Room Temperature
8.05
Molecular Biology Grade
Liquid
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.