missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v)

Catalog No. 15593031
Encompass
Change view
Click to view available options
Quantity:
100 mL
400 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15593031 100 mL
15593049 400 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15593031 Supplier Invitrogen™ Supplier No. 15593031

UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v) is used in the purification of nucleic acids. This reagent consists of highly pure chloroform, isoamyl alcohol, and UltraPure™ Phenol saturated with Tris-HCl.

  • Reagent consists of highly pure chloroform, isoamyl alcohol, and UltraPure™ Phenol saturated with Tris-HCl
  • When mixtures are extracted with Phenol:Chloroform:Isoamyl Alcohol, proteins are denatured and collected in the organic phase or at the interphase, while nucleic acids remain in the aqueous phase
  • UltraPure™ Phenol:Chloroform:lsoamyl Alcohol contains no preservatives
  • It is packaged under an inert gas in shatter-resistant, plastic-coated amber bottles
  • The appearance of the solution is evaluated at room temperature
  • No RNase or DNase activity is detected
  • Store in the refrigerator at 2°C to 8°C

Order Info

Shipping Condition: Room temperature

Specifications

Chemical Name or Material Phenols
Content And Storage Store at 2°C to 8°C. To extend product stability, it is recommended that the gas volume above the solution be flushed with argon or nitrogen following each use.
pH 8.05
Product Line UltraPure
Purity Molecular Biology Grade
Quantity 100 mL
Shipping Condition Room Temperature
Form Liquid
Do you have any information on DNA and RNA purification using phenol chloroform and alcohol precipitation?

Phenol extraction of proteins:

Phenol extraction is frequently used to remove proteins from nucleic acid solutions. A common protocol is to add an equal volume of buffer-saturated phenol or phenol:chloroform:isoamyl alcohol (25:24:1, v/v/v) to an aqueous nucleic acid solution, vortex, and centrifuge at 14,000 x g for 1 min to separate the phases.

Studies at Thermo Fisher Scientific have shown that the concentration of NaCl in the aqueous solution should not exceed 0.5 M for good recovery of DNA. Residual phenol can be removed from the aqueous phase by extraction with an equal volume of chloroform or ether. After extraction, DNA is usually precipitated with ammonium acetate and ethanol as described in another protocol on this server. Ref. Karger, B. D. (1989) FOCUS 11, 14.

A good source of general information on the properties of phenol can be found in Wallace, Donald M. “Large and Small-Scale Phenol Extractions”. Methods in Enz. Volume 152 guide to Molecular Cloning Techniques. 1987. Academic Press, Inc. Berger and Kimmel, eds. Chap.4, pg 33-41.

(a) At pH 5 to 6 DNA is selectively retained in the organic phase and interphase, leaving RNA in the aqueous phase. Therefore a pH greater than 7 is needed if DNA is to be extracted.

(b) At pH values below 7.6, poly A+ RNA is lost to the organic phase if chloroform is not present.

(c) Optimal RNA yields in phenol extraction are obtained if the salt concentration is less than 0.15 M NaCl. Salt concentration in the sample is not a factor for larger DNA molecules.

To store RNA after extraction use DEPC-treated water.

What is the recommended protocol for phenol-extraction removal of proteins from nucleic acid containing solutions?

Below is a commonly used protocol:

(1) Add an equal volume of buffer-saturated phenol or phenol:chloroform:isoamyl alcohol (25:24:1, v/v/v) to an aqueous nucleic acid solution. Note: for RNA solutions, acid-phenol is recommended.

(2) Vortex, and centrifuge at 14,000 x g for 1 min to separate the phases.

(3) Residual phenol can be removed from the aqueous phase by extraction with an equal volume of chloroform or ether.

(4) After extraction, DNA/RNA is usually precipitated with ammonium acetate and ethanol.

When using a proofreading polymerase such as Pfx, ThermalAce, Pfu, or Vent, is it necessary to neutralize or remove the enzyme after regenerating A-overhangs with a Taq polymerase?

It is best practice to remove the enzymes via phenol-chloroform extraction to prevent the proofreading enzyme from removing the A-overhang again after Taq incubation. If not done, TA cloning efficiencies could be 4-10 fold lower. Alternatively, the PCR product can be cleaned up by gel purification or PCR cleanup column.

What is the optimal pH of the phenol:chloroform mixture for isolation of DNA?

Partitioning of the nucleic acids in phenol is pH dependent. At pH 7.0 or higher, both DNA and RNA partition into the aqueous phase. At an acidic pH (below 7.0) DNA is denatured and will move into the organic phase, but the RNA remains in the aqueous phase. The mixture should be adjusted to at least pH 7.4 for work with DNA.

Recently I came across a DNA purification technique, which uses urea during phenol extraction. What is the purpose of using urea?

Using urea during phenol extraction denatures the protein associated with the DNA and the proteins that bind the genomic DNA to the cell wall.

For Research Use Only. Not for use in diagnostic procedures.