missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Glycine

Catalog No. 15527013
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15527013 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15527013 Supplier Invitrogen™ Supplier No. 15527013

Suitable for buffering at extremes of common buffering range

  • UltraPure Glycine is amino acid, with pKa1 of 2.3 and a pKa2 of 9.6, used in preparation of some electrophoresis buffers

1D Gel Electrophoresis, ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), Protein Gel Electrophoresis, Proteins, Expression, Isolation & Analysis, RNAi, Epigenetics & Non-Coding RNA Research

Order Info

Shipping Condition: Approved for shipment at room temperature or on wet ice

Specifications

Chemical Name or Material Glycine
Content And Storage Store at 15°C to 30°C.
Product Line UltraPure
Product Type Glycine
Quantity 500 g
Purity Molecular Biology Grade
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Powder

For Research Use Only. Not for use in diagnostic procedures.