missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ One-Cycle cDNA Kit for use with Thermo Scientific™ One-Cycle Assays

Catalog No. A10752030
Encompass
Change view
Click to view available options
Quantity:
30 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10752030 30 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10752030 Supplier Invitrogen™ Supplier No. A10752030

Includes

50μM T7-Oligo(dT) Primer, 5X 1st Strand Reaction Mix, 0.1M DTT, 10mM dNTP, SuperScript™ II (200u/μL), 5X 2nd Strand Reaction Mix, E.Coli DNA Ligase, E.Coli DNA Polymerase I, RNase H, T4 DNA Polymerase, 0.5M EDTA, RNase-free Water

Conveniently contains all of the reagents required for double-stranded cDNA synthesis within the GeneChip™ workflow

The kit includes Invitrogen's proprietary RNase H reduced activity SuperScript™ II Reverse Transcriptase and a qualified oligo (dt)-containing primer to perform reactions standard to Affymetrix– protocol. Designed to consistently produce higher yields of full-length cDNA for Affymetrix One-Cycle assays using either total RNA or poly A+ selected RNA (mRNA).

Order Info

Shipping Condition: Dry Ice

Specifications

Content And Storage 50 μM T7-Oligo(dT) Primer, 5X 1st Strand Reaction Mix, 0.1M DTT, 10 mM dNTP, SuperScript™ II (200u⁄ul), 5X 2nd Strand Reaction Mix, E.Coli DNA Ligase, E.Coli DNA Polymerase I, RNase H, T4 DNA Polymerase, 0.5M EDTA, RNase-free Water
All components are guaranteed stable for 6 months when properly stored.
Detection Method Biotin-based
Format Kit
Label or Dye Biotin
Final Product Type cDNA (Double-stranded)
Sample Type Poly(A+) RNA, Total RNA
Labeling Target cDNA
For Use With (Application) For generating double-stranded cDNA
Includes Label or Dye No
For Use With (Equipment) Affymetrix GeneChip™
Labeling Method Direct Labeling
Product Line SuperScript
Product Type cDNA Kit
Quantity 30 Reactions
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.