Learn More
Invitrogen™ PureLink™ PCR Micro Kit
PureLink PCR Micro Kits are ideal for purifying and concentrating DNA from low-yielding PCR or restriction digestion reactions.
Specifications
| Content And Storage | Kit includes columns, binding and wash buffers, recovery tubes, and the manual. Store at room temperature. |
|---|---|
| Format | Spin Column |
| Elution Volume Max | 20 μL |
| Elution Volume Min | 5 μL |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
Includes: Columns, binding and wash buffers, recovery tubes, and the manual
Description
PureLink™ PCR Micro Kits are ideal for purifying and concentrating DNA from low-yielding PCR or restriction digestion reactions.These highly purified DNA fragments and amplicons provide excellent results in downstream applications such as DNA sequencing, restriction digestion, cloning, PCR, and labeling. PureLink™ PCR Micro Kits provide a simple and rapid method for purifying and concentrating DNA fragments ranging in size from 100 bp-12 kb.
The unique micro-nozzle design allows up to 95% recovery of DNA fragments with as little as a 5μL elution volume. With less than a 10% coefficient of variance in the elution volume you can be sure that no ethanol is carried over from the wash steps giving you the best chance of success in downstream applications. Removal of primers, dNTPs, enzymes, and salts exceeds 98.5%. This kit can also efficiently remove unincorporated dye-labeled nucleotides from a cDNA labeling reaction.
Not High Throughput-Compatible (Manual)
Order Info
Shipping Condition: Room Temperature
Specifications
| Kit includes columns, binding and wash buffers, recovery tubes, and the manual. Store at room temperature. | |
| 20 μL | |
| Not High-throughput Compatible (Manual) | |
| PCR Micro Kit | |
| 100 bp to 12 kb | |
| 125 μL | |
| Cloning, Standard PCR |
| Spin Column | |
| 5 μL | |
| PureLink | |
| 10 min. | |
| Room Temperature | |
| DNA from Enzymatic Reactions (e.g. PCR) | |
| Silica Spin Column |
For Research Use Only. Not for use in diagnostic procedures.