Learn More
Invitrogen™ SYBR GreenER™ qPCR SuperMix for iCycler™ Instrument
Incorporate the latest high-performance technology to provide the most reliable gene expression data from your iCycler iQ™ or MyiQ™ instrument.
$644.00 - $10,270.00
Specifications
| Concentration | 2X |
|---|---|
| Content And Storage | 2X SYBR™ GreenER™ qPCR SuperMix for iCycler™ instrument contains: hot-start Taq DNA polymerase, SYBR™ GreenER™ fluorescent dye, fluorescein, MgCl2, dNTPs (with dUTP instead of dTTP), UDG, and stabilizers. Store at 4°C upon receipt. Guaranteed stable for 6 months when properly stored. |
| Detection Method | SYBR |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
Includes: Hot-start Taq DNA polymerase, SYBR GreenER fluorescent dye, fluorescein, MgCl2, dNTPs (with dUTP instead of dTTP), UDG, and stabilizers
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
11761100
|
Invitrogen™
11761100 |
100 Reactions |
Each for $644.00
|
|
|||||
11761500
|
Invitrogen™
11761500 |
500 Reactions |
Each for $2,910.00
|
|
|||||
1176102K
|
Invitrogen™
1176102K |
2000 Reactions |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The SYBR™ GreenER™ qPCR SuperMix kits include a dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range.
Consistent Specificity and Low-Copy Detection: The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets. The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity. With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA.
- The SYBR™ GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility
- A novel dsDNA-binding dye - exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR™ Green I, while using the same instrument filters and settings
- An optimized buffer system - improves sensitivity and provides excellent long-term stability
- Uracil DNA glycosylase - reduces carryover contamination in qPCR
Order Info
Shipping Conditions: Dry Ice
Specifications
| 2X | |
| SYBR | |
| qPCR | |
| Standard | |
| Platinum, SYBR GreenER | |
| Gene Expression |
| 2X SYBR™ GreenER™ qPCR SuperMix for iCycler™ instrument contains: hot-start Taq DNA polymerase, SYBR™ GreenER™ fluorescent dye, fluorescein, MgCl2, dNTPs (with dUTP instead of dTTP), UDG, and stabilizers. Store at 4°C upon receipt. Guaranteed stable for 6 months when properly stored. |
|
| High | |
| Taq DNA Polymerase | |
| BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5 | |
| Real Time PCR SYBR Master Mix | |
| DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.