missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR GreenER™ qPCR SuperMix for iCycler™ Instrument

Incorporate the latest high-performance technology to provide the most reliable gene expression data from your iCycler iQ™ or MyiQ™ instrument.

Supplier:  Invitrogen™ 11761500

Encompass

 View more versions of this product

Catalog No. 11761500


Only null left

Includes: Hot-start Taq DNA polymerase, SYBR GreenER fluorescent dye, fluorescein, MgCl2, dNTPs (with dUTP instead of dTTP), UDG, and stabilizers

Description

Description

The SYBR™ GreenER™ qPCR SuperMix kits include a dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range.

Consistent Specificity and Low-Copy Detection: The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets. The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity. With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA.

  • The SYBR™ GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility
  • A novel dsDNA-binding dye - exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR™ Green I, while using the same instrument filters and settings
  • An optimized buffer system - improves sensitivity and provides excellent long-term stability
  • Uracil DNA glycosylase - reduces carryover contamination in qPCR

Order Info

Shipping Conditions: Dry Ice

Specifications

Chemical Identifiers

2X
SYBR
qPCR
Standard
Platinum, SYBR GreenER
Gene Expression
2X SYBR™ GreenER™ qPCR SuperMix for iCycler™ instrument contains: hot-start Taq DNA polymerase,
SYBR™ GreenER™ fluorescent dye, fluorescein, MgCl2, dNTPs (with dUTP instead of dTTP), UDG, and stabilizers.

Store at 4°C upon receipt. Guaranteed stable for 6 months when properly stored.
High
Taq DNA Polymerase
BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5
Real Time PCR SYBR Master Mix
DNA (Genomic), cDNA

Specifications

2X
SYBR
qPCR
Standard
Platinum, SYBR GreenER
500 reactions
500 Reactions
500 Reactions
2X SYBR™ GreenER™ qPCR SuperMix for iCycler™ instrument contains: hot-start Taq DNA polymerase,
SYBR™ GreenER™ fluorescent dye, fluorescein, MgCl2, dNTPs (with dUTP instead of dTTP), UDG, and stabilizers.

Store at 4°C upon receipt. Guaranteed stable for 6 months when properly stored.
High
Taq DNA Polymerase
BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5
Real Time PCR SYBR Master Mix
Dry Ice
Gene Expression
DNA (Genomic), cDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.