missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ SYBR™ Green qPCR SuperMix-UDG w/ROX DFS Item

Ready-to-use reaction mixes for high-specificity, real-time amplification of cDNA, genomic DNA or plasmid DNA.

$726.00 - $2835.00

Specifications

Concentration 2X
Detection Method SYBR
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
View More Specs

Includes: 2X SuperMix with SYBR Green I and ROX Reference Dyes, and 50mM MgCl2

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity  
Catalog Number Mfr. No. No. of Reactions Price Quantity  
11744100 DFS Item
View Documents Promotion Details
Invitrogen™
11744100
100 Reactions
Each for $726.00
Only null left
 
11744500 DFS Item
View Documents Promotion Details
Invitrogen™
11744500
500 Reactions
Each for $2,835.00
Only null left
 
Description

Description

Platinum™ Taq DNA Polymerase activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification. Uracil-DNA Glycosylase (UDG) prevents carryover contamination in conjunction with dUTP (included) for improved specificity. ROX reference dye is included in the mix for maximum convenience. The ROX reference dye premixed at optimal concentration for ABI instruments.

Platinum™ SYBR™ Green qPCR SuperMix-UDG with ROX provides a sensitive, cost-effective system for real-time PCR detection with SYBR™ Green I dye

Order Info

Shipping Conditions: Dry Ice

Specifications

Specifications

2X
High
Taq DNA Polymerase
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System
Real Time PCR SYBR Master Mix
Gene Expression
SYBR
qPCR
Standard
Platinum, SYBR
Dry Ice
DNA (Genomic), cDNA
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.