Learn More
Invitrogen™ Platinum™ SYBR™ Green qPCR SuperMix-UDG w/ROX
Ready-to-use reaction mixes for high-specificity, real-time amplification of cDNA, genomic DNA or plasmid DNA.
Supplier: Invitrogen™ 11744500
Description
Platinum™ Taq DNA Polymerase activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification. Uracil-DNA Glycosylase (UDG) prevents carryover contamination in conjunction with dUTP (included) for improved specificity. ROX reference dye is included in the mix for maximum convenience. The ROX reference dye premixed at optimal concentration for ABI instruments.
Platinum™ SYBR™ Green qPCR SuperMix-UDG with ROX provides a sensitive, cost-effective system for real-time PCR detection with SYBR™ Green I dyeOrder Info
Shipping Conditions: Dry Ice
Specifications
| 2X | |
| SYBR | |
| qPCR | |
| Standard | |
| Platinum, SYBR | |
| 500 reactions | |
| 500 Reactions of 50 μL Each | |
| 500 Reactions | |
| 12.5 mL |
| Contains 2X Platinum SYBR Green qPCR SuperMix-UDG with ROX (12.5 mL) and 50 mM Magnesium Chloride (2 × 1 mL). Contains sufficient reagents for 500 reactions of 50 μL each. Store in freezer (-5 to -30°C). |
|
| High | |
| Taq DNA Polymerase | |
| 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System | |
| Real Time PCR SYBR Master Mix | |
| Dry Ice | |
| Gene Expression | |
| DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.