Learn More
Invitrogen™ Taq DNA Polymerase, native
Taq DNA Polymerase is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer.
Supplier: Invitrogen™ 18038067
Includes: Contains: 20μL Taq DNA Polymerase (5U/μL); 1.25mL 10X PCR buffer (without magnesium); 1mL Magnesium Chloride (50mM)
Description
Taq DNA Polymerase, native, is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer. This native enzyme is purified from Thermus aquaticus YT1 and consists of a single polypeptide with a molecular weight of 94 kDa. It has a 5' to 3' DNA polymerase activity and a 5' to 3' exonuclease activity.
Features
- A choice of recombinant or native enzyme
- Amplification of PCR products up to 5 kb in size
- Enzyme is licensed and qualified for PCR
Applications
- Amplification of DNA from complex genomic, viral, and plasmid templates
- RT-PCR
- Sequencing ssDNA
- Cycle sequencing
Notes
- One unit of Taq DNA Polymerase is the amount of enzyme required to incorporate 10 nmol of deoxyribonucleotide into DNA in 30 min. at 74°C.
Order Info
Shipping Condition: Approved for shipment on wet or dry ice
Chemical Identifiers
| 5 U/μL | |
| Low | |
| Standard | |
| DNA Polymerase | |
| Standard PCR | |
| No | |
| Separate Components | |
| DNA |
| Stand-alone enzyme | |
| Taq DNA Polymerase | |
| 5' - 3' | |
| Wet or Dry Ice | |
| 1X | |
| 3'-A | |
| 5 kb or less |
Specifications
| 5 U/μL | |
| Stand-alone enzyme | |
| qPCR, Standard PCR | |
| Standard | |
| DNA Polymerase | |
| Wet or Dry Ice | |
| 1X | |
| 1500 Reactions | |
| Separate Components | |
| DNA |
| • Taq DNA Polymerase (5 U/μL), 300 μL • 10X PCR buffer (without magnesium), 7.5 mL • 50 mM MgCl2, 3 mL Store at -20°C in a non-frost-free freezer. Stable for 6 months when stored properly. |
|
| Low | |
| Taq DNA Polymerase | |
| 5' - 3' | |
| 3 x 500 Units | |
| Standard PCR | |
| No | |
| 3'-A | |
| 5 kb or less |
For Research Use Only. Not for use in diagnostic procedures.