missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mimic™ Sf9 Insect Cells

Catalog No. 12552014
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12552014 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12552014 Supplier Gibco™ Supplier No. 12552014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Modified to stably express a variety of mammalian glycosyltransferases

Mimic Sf9 Insect Cells are a derivative of the Sf9 insect cell line. Typically, insect cells are unable to process N-glycans to the extent that mammalian cells do. This can affect protein structure, function, antigenicity, and enzymatic activity. The addition of mammalian glycosyltransferases to the Mimic™ Sf9 Insect Cells allows for production of biantennary, terminally sialyated N-glycans from insect cells. The cells can be used to produce more mammalian-like proteins in both baculovirus and stable insect expression systems.

1mL of Mimic Sf9 Insect Cells are provided frozen in 60% complete TNM-FH, 30% FBS, 10% DMSO at a concentration of 107 cells/mL.

Bioproduction, Cell Culture, Insect Cell Culture, Insect Expression, Insect Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Sf9 and Sf21 Cell Culture, Viral Vaccine Production

Specifications

Content And Storage Store in liquid nitrogen.
Cell Line Sf9
Cell Type Insect Cells
Species S. frugiperda
Product Line Mimic
Product Type Insect Cells
Quantity 1 mL
What is the procedure to thaw frozen insect cells?

The following protocol describes a general procedure for thawing cryopreserved cells. For detailed protocols, always refer to the cell-specific product insert.

1. Remove the cryovial containing the frozen cells from liquid nitrogen storage and immediately place it into a 37°C water bath.
2. Quickly thaw the cells (< 1 minute) by gently swirling the vial in the 37°C water bath until there is just a small bit of ice left in the vial.
3. Transfer the vial into a laminar flow hood. Before opening, wipe the outside of the vial with 70% ethanol.
4. Transfer the desired amount of pre-warmed complete growth medium appropriate for your cell line dropwise into the centrifuge tube containing the thawed cells.
5. Centrifuge the cell suspension at approximately 200 x g for 5-10 minutes. The actual centrifugation speed and duration varies depending on the cell type.
6. After the centrifugation, check the clarity of supernatant and visibility of a complete pellet. Aseptically decant the supernatant without disturbing the cell pellet.
7. Gently resuspend the cells in complete growth medium, and transfer them into the appropriate culture vessel and into the recommended culture environment.

Note: The appropriate flask size depends on the number of cells frozen in the cryovial, and the culture environment varies based on the cell and media type.

Why does the Insect cell line manual state: "Cells should be maintained at 27 degrees C in a non-humidified environment."

Insect cells do not require CO2 or high humidity to grow, they can grow in a lab drawer at room temperature. We recommend this so people don't waste CO2 and other resources necessary for maintaining a tissue culture incubator. It should be noted, however, that the cells will grow in a humidified incubator.

How can I concentrate my insect cells to increase the cell density?

If the cell density is too low and the cells have been in culture for 4-5 days, we recommend concentrating the cells by centrifuging them at 100 X g for 5 minutes and resuspending them in fresh medium. Cells should not be left in the same medium for more than 4-5 days as nutrients in the medium will have been used up by the cells in that period, and the medium itself degraded due to prolonged exposure to warm temperatures. Cells should also be centrifuged and concentrated if a lot of cell debris is observed in culture.

What are the main differences between insect cell culture and mammalian cell culture?

Insect cells are much more fragile than a lot of mammalian cell lines. They suffer much more damage than mammalian cells from overgrowth and over-splitting. Never let cells go above 8 x 10E6 cells/mL or grow at densities less than 0.5 x 10E6 cells/mL in suspension. Insect cells require a little more osmotic pressure than mammalian cells (340 µOsM). Insect cells use a lot of O2, especially during protein expression. Insect cell culture media is more acidic than mammalian media (pH 6.0-6.4). The insect cell culture media is phosphate buffer based. Therefore, no CO2 is needed to maintain the pH.

Which insect cell line is best to use?

We recommend Sf9 or Sf21 cells for transfection, purification, and amplification of recombinant virus. Sf9 cells are regular in size, easy to manipulate, and form good monolayers for plaque assays. Sf9 and Sf21 cells can also be used for expression of recombinant proteins, but the High Five cell line may produce higher levels. We recommend the High Five cell line for expression of secreted recombinant proteins. They are grown in serum-free medium, adaptable to suspension culture, and produce high levels of recombinant protein (Davis et al., 1992; see http://www.ncbi.nlm.nih.gov/pubmed/1368794).

Note: Generally it is easier to use one cell line for procedures up to optimization of protein expression. Once you have confirmed expression of your recombinant protein, other cell lines can be tried for optimization of expression levels.

Can I grow insect cells for short periods at 37 degrees C?

No, this is not recommended. Prolonged exposure to temperature higher than 29 degrees C will cause cell death. It is better to grow the cells at 27 degrees C or room temperature.

Is CO2 required for insect cell culture?

No, CO2 exchange is not required for insect cell culture.

What is the best way to plate insect cells?

Prior to performing transfections and plaque assays, cells need to be evenly distributed over the surface of a tissue culture plate. This ensures that:
- Cells do not distribute unevenly, leading to asymmetric monolayers.
- Maximum cell surface area is available for infection.

To disperse cells:
1. Rock the flask or plate slowly by hand forward and backward, then side-to-side.
2. Repeat this four times, watching carefully to ensure that the liquid reaches all areas of the growth surface.
3. Do not use a circular motion to disperse the cells, because this causes a concentration of cells around the edges of the plate rather than an even distribution.

What are the doubling times for robust growth and maintenance of different insect cell lines?

Please see below for information related to the type of cells, media, and doubling time in hours:

D.mel-2; Schneider's Drosophila + 10% FBS heat-inactivated; 18 to 24
High Five cells; Express Five SFM; approximately 24
Sf9 cells; Sf-900 II SFM; 24 to 30
Sf9 cells; Sf-900 III SFM; 24 to 30
Sf21 cells; Sf-900 II SFM; 24 to 30
Sf21 cells Sf-900 III SFM; 24 to 30

What are the sizes of Sf9, Sf21, High Five cells, and D.mel-2 cells?

Please see below for information related to type of cells, media, and cell size in microns:

D.mel-2; Schneider's Drosophila + 10% FBS heat-inactivated; 10 to 12
High Five cells; Express Five SFM; 17.5 to 19.5
Sf9 cell; Sf-900 II SFM; 15 to 17.5
Sf9 cells; Sf-900 III SFM; 15 to 17.5
Sf21 cell;s Sf-900 II SFM; 15 to 17.5
Sf21 cells; Sf-900 III SFM; 15 to 17.5

Does the incubator need to be humidified when culturing insect cells?

Humidification is only necessary for monolayer cultures and suspension cultures with volumes less than 35 mL. With shaker or spinner cultures greater than 35 mL, humidification is not necessary.


For Research Use Only. Not for use in diagnostic procedures.