missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 50 mL Tube Magnetic Separator

Catalog No. CS15050
Encompass
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CS15050 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CS15050 Supplier Invitrogen™ Supplier No. CS15050
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

One Magnetic Separator

Provides effective magnetic strength from neodymium magnets, which are aligned with plate wells or tubes.

  • Compatible with volumes used in kit protocols
  • Provides effective magnetic strength from neodymium magnets, which are aligned with plate wells or tubes

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Contains one Magnetic Separator, which may be stored at room temperature.
For Use With (Equipment) MagnaRack™ Magnetic Separator, Magna-Sep™ Magnetic Particle Separator
Shipping Condition Room Temperature
Product Line MagnaRack
Quantity 1 Each
Type Magnetic Separator
Capacity (Metric) 50 mL
I used water to elute my plasmid instead of the Elution Buffer with my ChargeSwitch kit, but saw very low plasmid yield. Why is this?

We strongly recommend using the Elution Buffer provided in the kit, and do not recommend elution with water. If you need to elute in any other buffer, be sure to use a buffer of pH 8.5-9.0 for efficient DNA elution.

What is the difference between the ChargeSwitch Pro kit and the Charge-Switch Pro filter kit?

The filter kit is based upon a dual nested column design, which facilitates the fastest and simplest protocol for obtaining high quality plasmid (less than 10 EU/µg DNA). Modified wash buffer 1 removes more endotoxin.

What is the typical yield when using the ChargeSwitch Pro Filter Plasmid Mini or Midi Kits?

The typical yield is 25 µg from a 5 mL culture for the ChargeSwitch Pro mini kit, 200 µg from a 25 mL culture for the midi kit, and 800 µg from a 100 mL culture from the maxi kit. The plasmid size range for the kits is typically 3-9 kb.

How does ChargeSwitch technology work?

Based on a patented switchable chemistry, ChargeSwitch purification utilizes the charge on a solid surface for nucleic acid capture. At lower pH, the surface chemistry holds a positive charge and attracts nucleic acid. At higher pH, the surface chemistry is neutral and any bound nucleic acid is released.

For Research Use Only. Not for use in diagnostic procedures.