Learn More
Gibco™ Drosophila S2 Cells in Schneider's Medium
Description
Gibco Drosophila S2 cells are used for heterologous protein expression in the DrosophilaM Expression System (DES™). Many features of the S2 cell line suggest that it is derived from a macrophage-like lineage. S2 cells grow at room temperature in ambient CO2 as a loose, semi-adherent monolayer in tissue culture flasks and in suspension in spinners and shake flasks.
- Expression of protein transiently or through generation of stable cell lines
- Quality and performance testing
Cell Culture, Drosophila S2 Cell Culture, Insect Cell Culture, Insect Expression, Insect Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Content And Storage | Storage conditions: Liquid nitrogen (vapor phase) Shipping conditions: Frozen |
| Cell Line | S2 (Schneider 2) |
| Cell Type | Insect Cells |
| Species | Drosophila |
| Product Type | Drosophila S2 Cells |
| Quantity | 1 mL |
Frequently Asked Questions (FAQs)
When grown at the optimal temperature of 27-28ºC, Drosophila S2 cells grow very quickly, with an approximate 14-16 hr doubling time. The cells can also be grown at 22-25ºC with doubling time of 24hrs. Humidity is an issue, so a shallow pan of water can be left in the incubator, but the incubator does not need to be turned on.
Growth medium is also a factor in doubling time - the doubling times listed here are for Sf900 II medium.
The cells can reach maximum densities of 1 to 2.5 x 10E7 cells/ml in shake flask cultures.
CuSO4 is water soluble. You can dissolve it in water for cell culture use, and sterile-filter it.
For Research Use Only. Not for any animal or human therapeutic or diagnostic use.