missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Low Range Quantitative DNA Ladder
GREENER_CHOICE

Catalog No. 12373031
Encompass
Change view
Click to view available options
Quantity:
100 applications
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12373031 100 applications
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12373031 Supplier Invitrogen™ Supplier No. 12373031

Includes

The E-Gel™ Low Range Quantitative DNA Ladder is supplied at 17.5ng/μL in 8mM Tris-HCl (pH 7.5), 1mM EDTA, 5% glycerol, and 0.005% Orange G; For 100 applications

Suitable for estimating size and quantity of double-stranded DNA by ethidium bromide visualization on all E-Gel™

Prepared from a specifically constructed plasmid, the ladder consists of an equimolar mixture of five blunt-ended fragments ranging from 100 to 2000bp. This ladder is supplied ready to use in an optimized E-Gel loading buffer. The buffer includes Orange G dye that runs at approximately 25bp. Compatible with E-Gel™, E-Gel™ 48, E-Gel™ 96 gels.

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

Specifications

Content And Storage • 1 mL E-Gel Low Range Quantitative DNA Ladder
• 1 mL 1X E-Gel Sample Loading Buffer

Store at room temperature or at 4°C for up to 6 months.
For longer periods, store at -20°C.
Concentration 0.0175 μg/μL
Gel Type Agarose
Ready to Load Yes
Sample Loading Volume 1 mL
Size Range 100 to 2000 bp
Volume (Metric) 1000 μL
Gel Compatibility E-Gel™ agarose gels
Quantity 100 applications
Shipping Condition Approved for shipment at Room Temperature or on Dry Ice
Technology Individual chromatography-purified DNA fragments
Product Type DNA Ladder
Show More Show Less
Can I know the sequences of Invitrogen DNA ladders?

Sequences of Invitrogen DNA and RNA ladders are proprietary.

Are Invitrogen DNA ladders composed of linear or circular/supercoiled DNA?

Invitrogen DNA ladders contain linear dsDNA fragments.

Are Invitrogen DNA ladders composed of single-stranded or double-stranded DNA fragments?

Invitrogen DNA ladders are composed of double-stranded DNA fragments only.

I'm seeing anomalous migration of my DNA ladder. What happened?

This can happen if the marker was heated. Please ensure that the ladders are not heated before use.

What is the difference between the TrackIt DNA Ladders and other DNA ladders?

TrackIt DNA Ladders contain two tracking dyes in the sample buffers, which serve as visual markers for tracking electrophoresis progress through the gel, and also to indicate when maximum resolution is achieved. The tracking dyes should not obscure your visualization of DNA bands in the ladder, as the dyes run outside the limits of most DNA bands in the ladder.

TrackIt Cyan/Orange Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Orange G. We recommend TrackIt Cyan/Orange Loading Buffer for DNA fragments between 10 bp and 1 kb.

The TrackIt Cyan/Yellow Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Tartrazine. We recommend TrackIt Cyan/Yellow Loading Buffer for DNA fragments between 100 bp and 10 kb. The molecular weights are Xylene Cyanol FF, 638.6; Orange G, 452.4; Tartrazine, 534.4.

Note: The TrackIt DNA Ladders are not recommended for use with polyacrylamide gels and are not designed for quantitation.

I am using a quantitative DNA standard and see little difference between the bands when they are ethidium bromide stained. How can this occur and what can I do?

Here are a few suggestions:

(1) Excess volume loaded on gel. The smallest loading volume will result in the greatest difference in band thickness between different masses. Basically overloading the ladders gives saturated bands which do not show any variation in intensity. It is best to load all samples in the same volume.

(2) Sample loaded in larger or smaller volume than standard. Load equal volumes of sample and standard DNA for accurate comparison.

(3) Ethidium bromide (EtBr) staining after electrophoresis. Include ethidium bromide in the gel rather than staining after electrophoresis to increase the difference in band intensity.

(4) EtBr was in the gel, but not the running buffer. This results in the absence of EtBr in the lower half of the gel after electrophoresis and therefore differential fluorescence between bands in the upper half of the gel and the lower half of the gel.

What can cause anomalous migration of my DNA markers?

In general there are very few reasons why a DNA marker would run anomalously. Here are a few suggestions:

(1) For Lambda DNA/Hind III Fragments, the cos site reannealed. Heat Lambda DNA/Hind III Fragments at 65 degrees C for no longer than 10 min before electrophoresis.

(2) Improper electrophoresis conditions were used. Do not allow voltage to exceed ~20 V/cm. Maintain a temperature <30 degrees C during electrophoresis. Check that the electrophoresis buffer used has sufficient buffering capacity.

(3) DNA was denatured. Do not heat standards (except Lambda-derived markers) prior to electrophoresis. Dilute markers in a buffer containing 20 mM NaCl.

I ran my DNA molecular weight ladder, and several bands seem to be missing. Can you offer suggestions on how to rectify this?

There are a few possible reasons for this:

(1) Small DNA bands were electrophoresed off the gel. Electrophorese the gel for less time, at lower voltage, or use a higher percentage gel.

(2) The small bands migrated with the dye front due to differences in ionic strength between gel and buffer. Be sure the buffer in the gel is the same as the electrophoresis buffer.

(3) DNA bands of similar molecular size were not resolved. Increase the electrophoresis time and check the proper percentage gel for resolution. For DNA fragments <1,000 bp, try Agarose 1000 (Cat. No. 16550100) instead of agarose.

(4) DNA was denatured. Do not heat standards (except Lambda-derived markers) prior to electrophoresis. Dilute markers in a buffer containing 20 mM NaCl.

Are there any suggestions to improve the detection of the bands of my DNA molecular weight ladder in an agarose gel?

Faint or absent DNA ladders might be due to:

(1) An insufficient quantity or concentration of DNA loaded on the gel. You can increase the amount of DNA.

(2) Degraded DNA. Avoid nuclease contamination of the DNA standards. Use sterile or autoclaved tips.

(3) DNA was electrophoresed off the gel. Electrophorese the gel for less time, at a lower voltage, or use a higher percentage gel.

(4) An improper UV light source was used for ethidium bromide - stained DNA. Use short-wavelength (254 nm) UV light for greater sensitivity.

(5) The fluorescence bleached after long UV exposure. Stain the gel again with ethidium bromide.

(6) For Biotinylated Lambda DNA/Hind III Fragments or Biotinylated PhiX174/Hinf I Fragments, the biotin was removed by exposure to alkaline conditions. Minimize exposure of biotinylated DNA to NaOH. Note: Alkaline transfers to a membrane do not present a problem when performed at room temperature.

What could be causing smearing of my molecular weight DNA ladder?

(1) Incorrect concentration of DNA ladder gives poor separation and smearing of DNA bands.
(2) Samples containing ≥50 mM NaCl, 100 mM KCl, 10 mM acetate ions, or 10 mM EDTA (i.e. certain restriction enzyme and PCR buffers) will cause loss of resolution on E-Gel agarose gels.


For Research Use Only. Not for use in diagnostic procedures.