missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ EX Agarose Gels

Catalog No. G402022
Encompass
Change view
Click to view available options
Quantity:
10 Gels/Pk.
20 Gels/Pk.
Separation Range:
100 to 5000 bp
20 to 800 bp
50 to 2000 bp
Gel Percentage:
1%
2%
4%
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Separation Range Gel Percentage
G402022 20 Gels/Pk. 50 to 2000 bp 2%
G401001 10 Gels/Pk. 100 to 5000 bp 1%
G402021 20 Gels/Pk. 100 to 5000 bp 1%
G401002 10 Gels/Pk. 50 to 2000 bp 2%
G401004 10 Gels/Pk. 20 to 800 bp 4%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. G402022 Supplier Invitrogen™ Supplier No. G402022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Requires

Requires the E-Gel iBase™ Power System for running and the E-Gel Safe Imager™ Real-Time Transilluminator for real-time viewing of sample migration.

Invitrogen E-Gel EX precast agarose gels are designed for ultrasensitive and convenient DNA and RNA sample electrophoresis in as little as 10 to 20 minutes.

Invitrogen E-Gel EX precast agarose gels are designed for ultrasensitive and convenient DNA and RNA sample electrophoresis in as little as 10 to 20 minutes. Each E-Gel agarose gel cassette contains all components and stain required for efficient gel separation and analysis–just load your samples and run. E-Gel agarose gels are ideal for analyzing PCR products, restriction digests, and plasmid preparations, or for DNA fragment library analysis. E-Gel agarose gels are also suitable for a quick check of RNA samples.

Features of E-Gel EX agarose gels include:

  • Instantaneous setup—precast E-Gel agarose gels are ready for use when you are, and do not require gel preparation or additional buffers
  • High sensitivity—detect 1 ng of sample DNA
  • Rapid analysis—complete DNA sample separation in 10–20 minutes
  • In-process control—view DNA sample migration in real time
  • Easy sample access—openable cassette design allows easy in-gel access for convenient DNA sample excision or transfer to membranes
  • For DNA and RNA—ideal for DNA sample analysis and suitable for quick check of RNA samples

Ultra-sensitive detection of DNA

E-Gel EX gels pre-stained with SYBR Gold DNA stain were developed for ultimate sensitivity and demonstrate up to 5-fold greater sensitivity than comparable gels containing ethidium bromide. This superior sensitivity allows you to use lower amounts of sample, saving time and money.

Fast and convenient analysis

E-Gel EX gels are our fastest-resolving agarose gels, enabling sample analysis in as little as 10–20 minutes. Each E-Gel agarose gel contains agarose, electrodes, SYBR Gold DNA stain, and a bufferless ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not required any gel or buffer preparation or gel straining steps. Just load your samples and run.

Live monitoring of DNA migration

The E-Gel EX agarose gels are pre-stained with SYBR Gold II DNA gel stain with an excitation wavelength in the blue-light spectrum. When used on the E-Gel Power Snap Electrophoresis Device with integrated blue-light trans-illuminator, safe real-time monitoring of DNA or RNA migration is enabled without risk to eyes or possible damage of the sample, unlike UV illumination.

Quick check of RNA samples

Besides DNA analysis, E-Gel EX gels can also be used for fast analysis of RNA samples to check their integrity before proceeding with downstream applications.

Easy access to the gel

E-Gel cassettes are designed for convenient opening with a Gel Knife (Cat. No. EI9010), for easy excision of specific bands or transfer of the gel to a membrane for southern blot analysis.

Running and imaging devices

E-Gel EX gels require the Invitrogen E-Gel Power Snap Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.

Available in a variety of gel formats

E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The 11-well E-Gel EX agarose gels are available in 1%, 2%, and 4% gel percentages.

Order Info

Shipping Condition: Room Temperature

Specifications

Compatible Ladders E-Gel™ 1 Kb Plus Express Ladder, Century™-Plus RNA Markers
Content And Storage • E-Gel™ EX 20-Paks contain 20 gels

Store gels at room temperature. Guaranteed stable for 3 months when properly stored.
Gel Percentage 2%
Gel Type E-Gel
Label or Dye SYBR Gold
Sample Loading Volume 20 μL
Separation Range 50 to 2000 bp
Wells 11-well
Mode of Separation Molecular Weight
Quantity 20 Gels/Pk.
Sample Buffer Type Preparative without dyes, Non-preparative
Sample Type Double-Stranded DNA (dsDNA), Single-Stranded RNA, Circular RNA
Shipping Condition Room Temperature
Well Design Single Comb
Product Type EX Agarose Gel
Show More Show Less
What ladder do you recommend for running RNA on Invitrogen E-Gel EX Agarose Gels?

Depending on the RNA fragment size, we recommend the following ladders:

  • RiboRuler Low Range RNA ladder, (100 bp-1,000 bp) (Cat. No. SM1833)

  • RiboRuler High Range RNA ladder, (200 bp-6,000 bp) (Cat. No. SM1823)

Can you provide me with the protocol to run E-Gel EX gels for RNA samples on the E-Gel PowerSnap Electrophoresis System?

Yes, the protocol for running E-Gel EX gels for RNA samples on the E-Gel PowerSnap Electrophoresis System can be found in the E-Gel Power Snap Electrophoresis System User Guide, on page 46 under Appendix E.

Can E-Gel EX Agarose Gels, 2% be used in a standard gel electrophoresis apparatus?

No. E-Gel EX Agarose Gels, 2% are only compatible with the E-Gel Power Snap Electrophoresis System (Cat. No. G8300).

What are the size dimensions of E-Gel EX Agarose Gels, 2% (Cat. No. G402022)?

Please see page 22 of the E-Gel Technical Guide for size dimensions (https://tools.thermofisher.com/content/sfs/manuals/egelguide_man.pdf).

For Research Use Only. Not for use in diagnostic procedures.