missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Mouse CD8 (Lyt 2)

Catalog No. 11447D
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11447D 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11447D Supplier Invitrogen™ Supplier No. 11447D

Includes

5mL Dynabeads Mouse CD8 (Ly-2), coated with an anti-Ly2 monoclonal antibody. Lyt-2 antigen is expressed on thymocytes and the cytotoxic/suppressor subset of mature T cells from all common mouse strains

Dynabeads™ Mouse CD8 (Lyt 2) are magnetic beads that provide a robust and versatile tool to isolate or deplete mouse CD8+ T-cells directly from any sample.

Dynabeads™ Mouse CD8 (Lyt 2) are magnetic beads that provide a robust and versatile tool to isolate or deplete mouse CD8+ T-cells directly from any sample, including tissue digests such as spleen or lymph nodes, whole blood, bone marrow, and mononuclear cells (MNC).
• Rapid isolation of mouse CD8+ T-cells from any sample – no columns required
• Positive isolation for molecular downstream assays or efficient depletion of mouse CD8+ T cells

Excellent Yield, Purity, and Viability of Recovered Cells
Dynabeads™ Mouse CD8 (Lyt 2) are uniform, superparamagnetic beads (4.5 μm diameter) that enable easy isolation of mouse CD8+ cells from spleen or lymph nodes, although any single-cell suspension may be used. Dynabeads™ Mouse CD8 beads are coated with a primary monoclonal antibody specific for the Lyt 2 membrane antigen mainly expressed on cytotoxic/suppressor subpopulation of mature T-cells of all common mouse strains. Dynabeads™ Mouse CD8 binds to CD8+ cells during a short incubation period in a tube, after which the bead-bound cells are separated from unbound cells using a magnet. This fast and gentle isolation method does not require the use of columns, and helps ensure high purity, recovery and viability of the isolated CD8+ cells.

Ideal for Both Depletion and Positive Isolation for Downstream Molecular Assays
Because of the powerful magnets used for separation, Dynabeads™ Mouse CD8 (Lyt 2) can be used in viscous samples such as single cell suspensions from tissue, whole blood and bone marrow, and enable efficient depletion of mouse CD8+ cells in about 30 minutes. Dynabeads™ Mouse CD8 (Lyt 2) will also afford excellent recovery of high-purity, viable cells when used for positive isolation for downstream studies; for example, those in which cells are to be lysed while still attached to beads, and nucleic acids or proteins further purified. Note that intact cells will not be released from these beads, so if you want to isolate mouse CD8+ cells for cell-based applications, or you need to check your samples with flow cytometry, you should use Dynabeads™ FlowComp Mouse CD8 Cells (Cat. No. 11462D) (for bead-free cells).

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

Order Info

Shipping Condition: Room Temperature

Specifications

Cell Type T Cells (CD8)
Content And Storage This Product Contains: 5 mL Dynabeads™ Mouse CD8 (Ly-2), coated with an anti-Ly2 monoclonal antibody. Lyt-2 antigen is expressed on thymocytes and the cytotoxic/ suppressor subset of mature T cells from all common mouse strains.

Store at 2°C to 8°C.
Product Type Antibody Coated Bead
Isolation Technology Depletion, positive isolation for molecular applications
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 5 mL
Reactivity Mouse
Sample Type Lymph Node, Blood, Spleen
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 cells per isolation
Target Species Mouse
Diameter (Metric) 4.5 μm
Show More Show Less
My Dynabeads magnetic beads are not pelleting well with the magnet. Do you have any suggestions for me?

Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.

Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

I have a long double-stranded DNA fragment I would like to isolate. What product do you recommend?

For biotin-labled DNA that is less than 1 kb, we recommend you use Dynabeads M270 Streptavidin and MyOne C1 magnetic beads. We recommend our Dynabeads KilobaseBINDER Kit, which is designed to immobilize long (>1 kb) double-stranded DNA molecules. The KilobaseBINDER reagent consists of M-280 Streptavidin-coupled Dynabeads magnetic beads along with a patented immobilization activator in the binding solution to bind to long, biotinylated DNA molecules for isolation. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/immobilisation-of-long-biotinylated-dna-fragments.html) for more information in regards to long biotinylated DNA fragment isolation.

Can I use Dynabeads magnetic beads to isolate single-stranded DNA templates?

Yes, Dynabeads magnetic beads can be used to isolate single-stranded DNA. Streptavidin Dynabeads magnetic beads can be used to target biotinylated DNA fragments, followed by denaturation of the double-stranded DNA and removal of the non-biotinylated strand. The streptavidin-coupled Dynabeads magnetic beads will not inhibit any enzymatic activity. This enables further handling and manipulation of the bead-bound DNA directly on the solid phase. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/preparing-single-stranded-dna-templates.html) for more information in regards to single-stranded DNA capture.

What is the magnetic susceptibility for Dynabeads magnetic beads?

Magnetic susceptibility is a measure of how quickly the beads will migrate to the magnet. This will depend on the iron content and the character of the iron oxide. The magnetic susceptibility given for the Dynabeads magnetic beads is the mass susceptibility, given either as cgs units/g or m^3/kg (the latter being an SI unit). For ferri- and ferromagnetic substances, the magnetic mass susceptibility is dependent upon the magnetic field strength (H), as the magnetization of such substances is not a linear function of H but approaches a saturation value with increasing field. For that reason, the magnetic mass susceptibility of the Dynabeads magnetic beads is determined by a standardized procedure under fixed conditions. The magnetic mass susceptibility given in our catalog is thus the SI unit. Conversion from Gaussian (cgs, emu) units into SI units for magnetic mass susceptibility is achieved by multiplying the Gaussian factor (emu/g or cgs/g) by 4 pi x 10^-3. The resulting unit is also called the rationalized magnetic mass susceptibility, which should be distinguished from the (SI) dimensionless magnetic susceptibility unit. In general, magnetic mass susceptibility is a measure of the force (Fz) influencing an object positioned in a nonhomogenous magnetic field. The magnetic mass susceptibility of the Dynabeads magnetic beads is measured by weighing a sample, and then subjecting the sample to a magnetic field of known strength. The weight (F1) is then measured, and compared to the weight of the sample when the magnetic field is turned off (F0). The susceptibility is then calculated as K x 10^-3 = [(F1-F0) x m x 0.335 x 10^6], where K is the mass susceptibility of the sample of mass m. The susceptibility is then converted to SI units.

How can I determine coupling efficiency of Dynabeads magnetic beads?

There are different methods to check binding of ligands to the beads, including optical density (OD) measurement, fluorescent labeling, and radioactive labeling.

For OD measurement, you would measure the OD of the ligand before immobilization to the beads and compare it with the ligand concentration that is left in the supernatant after coating. This gives a crude measurement of how much protein has bound to the beads.

Protocol:

1.Set spectrophotometer to the right wavelength. As a blank, use the Coupling Buffer.
2.Measure the absorbance of the Pre-Coupling Solution. A further dilution may be necessary to read the absorbance, depending upon the amount of ligand added.
3.Measure the absorbance of the Post-Coupling Solution. A dilution may be necessary to read the absorbance.
4.Calculate the coupling efficiency, expressed as the % protein uptake, as follows. [(Pre-Coupling Solution x D) - (Post-Coupling Solution x D)] x 100/(Pre-Coupling Solution x D) where D = dilution factor.

For fluorescent labeling, we suggest negatively quantifying the amount of ligand bound by measuring ligand remaining in the coupling supernatant (compared to the original sample), rather than directly measuring the ligands on the beads. Add labeled ligand to the beads, and measure how much ligand is left in the supernatant (not bound to the beads). By comparing this with the total amount added in the first place, you can then calculate how much of the ligand that has been bound to the beads. Keep in mind that the Dynabeads magnetic beads are also autofluorescent, which is why direct measuring of fluorescence of the bead-bound ligands is not recommended, but rather this indirect approach. The label could be, for example, FITC/PE. Some researchers perform a direct approach with success (using a flow cytometer).

Radioactive labeling is the most sensitive method of the three, but it is also the most difficult one. It involves radioactively labeling a portion of the ligand. We use radiolabeled I-125 in tracer amounts and mix it with "cold" ligands in a known ratio before coupling. The absolute quantities for the ligand on the beads should be obtained by measuring the beads in a scintillation (gamma) counter and comparing the cpm with a standard.

Protocol:

1.Take out an appropriate amount of beads and wash the beads in 1 mL of binding buffer.
2.Pipette out desired amount of human IgG in a separate tube.
3.Mix the human IgG with I-125-labeled human IgG (30,000 - 100,000 cpm).
4.Dilute the mixture of human IgG and I-125-labeled human IgG to 100 mL in binding buffer.
5.Incubate for 30 minutes at room temperature and measure the cpm in a scintillation counter.
6.Wash the beads (with coating) four times, and measure cpm again.
The % binding is calculated by using the equation : (cpm after washing/cpm before washing)x100%.

What sizes do you offer for the Dynabeads magnetic beads?

Dynabeads magnetic beads come in three sizes: 4.5 µm (M-450), 2.8 µm (M-270/M-280), and 1 µm (MyOne beads). The largest of the Dynabeads magnetic beads is ideal for big targets like cells. The 2.8 µm beads are recommended for proteomics and molecular applications. The smallest of the beads, 1 µm, are ideal for automated handling.

Can Dynabeads magnetic beads be sonicated?

In general, short sonication is a good way to reduce aggregation of the beads and ensure optimal homogenous conditions at the time of ligand addition when coating the beads. When target is bound to the beads, more care is needed, as the binding might break. The streptavidin beads themselves should tolerate sonication. We have not tested sonication for long periods, but 5 minutes is fine. We do not have information about the streptavidin-biotin interaction being broken by such treatment.

Can Dynabeads magnetic beads be sterilized?

If desired, the uncoated epoxy or tosylactivated beads can be sterilized by washing with 70% ethanol. Coated beads cannot be sterilized.

What are Dynabeads magnetic beads?

Dynabeads magnetic beads are uniform, non-porous, superparamagnetic, monodispersed and highly cross-linked polystyrene microspheres consisting of an even dispersion of magnetic material throughout the bead. The magnetic material within the Dynabeads magnetic beads consists of a mixture of maghemite (gamma-Fe2O3) and magnetite (Fe3O4). The iron content (Fe) of the beads is 12% by weight in Dynabeads magnetic beads M-280 and 20% by weight in Dynabeads magnetic beads M-450. The Dynabeads magnetic beads are coated with a thin polystyrene shell which encases the magnetic material, and prevents any leakage from the beads or trapping of ligands in the bead interior. The shell also protects the target from exposure to iron while providing a defined surface area for the adsorption or coupling of various molecules.

Uniformity of bead size and shape provides consistent physical and chemical properties. These uniform physical characteristics lead to high-quality, reproducible results.

The Dynabeads magnetic beads are available in three different sizes: 4.5 µm (M-450 beads), 2.8 µm (M-270/M-280 beads) and 1 µm (MyOne beads). as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

Can I store the antibody/ligand-coated Dynabeads magnetic beads?

Dynabeads magnetic beads coated with antibody/ligand may be stored at 2-8 degrees C without loss of antigen binding capacity. For long-term storage, a final concentration of 0.02% NaN3 may be added to the antibody-coupled beads in a physiological buffer. Please note that not all coupled antibodies retain their function in long term storage. Verify your coupled antibody stability by testing in small scale. After storage, coated Dynabeads magnetic beads should be washed once in PBS/BSA for 5 min before use.

When isolating cells with Dynabeads magnetic beads, what is more important: bead-to-target cell ratio or the concentration of beads in the bead/cell mixture?

Both bead-to-target cell ratio and the concentration of beads in the bead/cell mixture are important and should be considered. For example, when using the Dynabeads magnetic beads M-450 CD4 positive isolation or depletion kit, a 4:1 bead-to-target cell ratio should be maintained. To capture 95% of target cells for molecular applications, the bead concentration must always be 1 x 10e7 beads per milliliter of sample. To deplete 99% CD4 cells from the starting sample, the bead concentration must always be 2 x 10e7 beads per milliliter of sample. Please consult the package insert for recommended bead concentrations of each product.

What is the definition of superparamagnetic, and what does this mean for my cell isolation application with Dynabeads magnetic beads?

Superparamagnetic means that the Dynabeads magnetic beads exhibit magnetic properties when placed within a magnetic field, but have no residual magnetism when removed from the magnetic field.

This means that your targeted cells, proteins, or nucleic acids are only subjected to magnetic forces during the time the beads are on the magnet. The beads do not aggregate, but remain evenly dispersed in suspension.

Are the antibodies on your Dynabeads magnetic beads for cell isolation/activation/expansion covalently bound to the beads?

Yes. The antibodies are covalently bound and should be very stable in your applications.

What is the average shelf life of Dynabeads magnetic beads?

Depending on the antibody coated on the Dynabeads magnetic beads, the shelf life can vary from 24-36 months.
Some kits may have 18 months shelf life depending on other components supplied in the kit. The kits are guaranteed for 6 months from when you receive them. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What do the designations M-280, M-270, and MyOne mean on Dynabeads magnetic beads?

The M stands for magnetic. M-280 refers to hydrophobic 2.8 micron beads, while M-270 refers to hydrophilic 2.8 micron beads. MyOne refers to 1 micron beads. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What is the detection limit when using Dynabeads magnetic beads for immunoprecipitation (IP)?

Answering this question is not straightforward. It will depend on the detection method. When using HRP (horseradish peroxidase)-based detection system or radioactivity in combination with a good antibody, very little target is required. More target is required when using an AP (alkaline phosphatase)-based detection system. When a sensitive detection system is used, detection will most likely be in the nanogram range. In some cases, pictograms of target can be detected.

What is the elution volume when using Dynabeads magnetic beads for immunoprecipitation (IP)?

Within practical limits, the elution volume can be scaled up or down to suit your experiment. However, volumes less than 10 µL become more difficult to work with. In addition, the amount of target is important. If you have a lot of beads with a lot of bound target in a small elution volume, your elution may not be very efficient. Typically, 15-100 µL of beads may be eluted in 30 µL. For efficient recovery of the antigen and/or binding partners, the elution volume should at minimum equal the volume of the beads.

How can I quantify the amount of antibody bound to Dynabeads magnetic beads?

There are several methods to quantify the amount of antibody bound to the beads. The crudest method is to measure the concentration of antibody in the coupling reaction before and after antibody attachment. Either fluorescence measurements or absorbance at 280 nm can be used. Alternatively, you could measure the amount of antibody bound to the beads by fluorescence, chemiluminescence, or radiolabeling detection methods. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

How long should I incubate my antibody with the lysates?

Incubation time will depend on the immunogenicity of the primary antibody and its binding affinity with the specific antigens. For a good primary antibody, 30-40 minutes incubation should work well. If you are working with a poor antibody or a very low-abundance protein, you could try to increase binding by incubating overnight. However, this also increases the chance of background protein binding.

When should I covalently bind the antibody to the Dynabeads surface?

If the target protein has the same molecular weight as the heavy or light chain antibody, we would recommend covalently binding the antibody to the bead surface. This can be done by either crosslinking the antibody to the Dynabeads Protein A or G magnetic beads, or secondary coated beads, or by using one of the surface-activated Dynabeads magnetic beads. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What are the general advantages of using Dynabeads magnetic beads for protein isolation?

Using Dynabeads magnetic beads for protein isolation provides several advantages:

-Rapid binding kinetics: since the number of beads per volume for Dynabeads is approximately 1,000 times higher than for the same volume of a Sepharose slurry, the probability for Dynabeads magnetic beads to hit the target is far greater.

-Incubation time: due to the rapid binding kinetics, the protocol is usually very short.
-Low background: due to the rapid binding kinetics and the short incubation time, the background is also very low.
-Trapping of impurities: the beads offer no internal volume for binding or trapping of impurities.
-Low antibody consumption: this is because Dynabeads magnetic beads are nonporous, uniform superparamagnetic, monodispersed, highly crosslinked polystyrene microspheres consisting of an even dispersion of magnetic material throughout the bead. The beads are coated with a thin layer of a highly crosslinked polystyrene shell that encases the magnetic material and prevents any leakage from the beads or trapping of ligands in the bead interior. The outer layer also provides a defined surface area for the adsorption or coupling of various molecules such as antibodies. Uniformity of bead size and shape provide consistent physical and chemical properties. These uniform physical characteristics lead to high-quality, reproducible results.
-Reproducibility: due to easier practical handling, such as pipetting. No centrifugation steps or preclearing are required.

Are Dynabeads magnetic beads compatible with dithionite, DTT, and EDTA?

No. Not only is dithionite a reducing agent, but the strong affinity of the dithionite ion for bivalent and trivalent metal cations (M2+, M3+) allows it to enhance the solubility of iron, making it a chelating agent. As a result, the iron in the Dynabeads magnetic beads is reduced and pulled out when they are exposed to dithionite. The same is observed if Dynabeads magnetic beads are exposed to DTT and EDTA. With EDTA, we highly recommend checking the minimal amount of EDTA that your specific molecules would tolerate for binding to the Dynabeads, and if it will affect your specific application. For some applications, low concentrations of EDTA can be tolerated by Dynabeads. On the other hand, using 10 mM EDTA with heating affects the binding of biotin molecules to Dynabeads streptavidin. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

Are Dynabeads magnetic beads compatible with Urea?

Yes, they are compatible with 6-8 M Urea when used during post-coupling steps.

Are Dynabeads magnetic beads compatible with centrifugation?

Dynabeads magnetic beads, being magnetic in nature are really not designed to be centrifuged. That being said, the beads themselves are compact, as the pores in the polymer matrix are filled with magnetic material and coated with a final outer polymer shell that will further add to the rigidity of the beads. Hence, pressure should theoretically not be a problem for the beads themselves, but the force exerted by the beads on surrounding cells in the pellet may be detrimental to the cells. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What are the benefits of using magnetic beads in immunoprecipitation (IP) experiments?

Magnetic beads, unlike agarose beads, are solid and spherical, and antibody binding is limited to the surface of each bead. While magnetic beads do not have the advantage of a porous center to increase the binding capacity, they are significantly smaller than agarose beads (1 to 4 µm), which collectively gives them adequate surface area-to-volume ratios for optimum antibody binding.

High-power magnets are used to localize magnetic beads to the side of the incubation tube and out of the way to enable cell lysate aspiration without the risk of also aspirating immune complexes bound to the beads. Magnetic separation avoids centrifugation, which can break weak antibody-antigen binding and cause loss of target protein.

However, an issue with the use of magnetic beads is that bead size variations may prevent all beads from localizing to the magnet. Additionally, while immunoprecipitation using agarose beads only requires standard laboratory equipment, the use of magnetic beads for immunoprecipitation applications requires high-power magnetic equipment that can be cost-prohibitive. Read more about our Magnetic Immunoprecipitation Products (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/immunoprecipitation.html#products).

How do I convert the Dynabeads magnetic bead concentration from 1 mg beads/mL to actual bead numbers per milliliter?

This depends on the bead size. In general, for M-450 Dynabeads, 1 mg/mL beads is approximately equivalent to 1.3 X 10E7 beads/mL; for M-280 Dynabeads, 1 mg/mL beads is approximately equivalent to 6.7 X 10E7 beads/mL.

Will Dynabeads magnetic beads be internalized if cultured with the beads on?

Whether cells will internalize the Dynabeads magnetic beads during culture will depend on the cell type. Due to the bead size (usually 4.5µm in diameter) Dynabeads magnetic beads will not be internalized into the endocytic pathway e.g., via clathrin coated pits. The clathrin coated pits are typically not more than 500 nm in size, which is far too small for endocytosis of the beads. However, if cells with phagocytic activities (e.g., monocytes/macrophages) are present, the Dynabeads magnetic beads will be phagocytosed into the phagolysosomes by these specialized cells. So it would really depend on the cell type.

Is there a method to remove the Dynabeads magnetic beads from isolated cells if the bead releasing reagent is not available?

We offer several Dynabeads magnetic beads that can be used for either positive isolation (keep the target cells) or for depletion (remove the target cell from a sample) that does not include any release mechanism:

- Dynabeads magnetic beads for depletion: Using Dynabeads magnetic beads for depletion is a very fast, efficient and easy method. Use pre-coated Dynabeads magnetic beads or coat your own target antibody onto our secondary coated beads, add to any sample (e.g., whole blood, PBMC, buffy coat, tissue digests), incubate for 20 minutes with mixing, apply to a magnet for 2 minutes, and you have your cells depleted.

- Dynabeads magnetic beads for positive isolation for molecular downstream assays: Positive isolation of target cells without bead release can be used when the aim is downstream molecular studies such as DNA, RNA, or protein analysis. In these applications, the isolated cells can be lysed while the beads are attached to the cells, and the beads can be removed after cell lysis. If the bead presence is not a problem, you can also culture the cells with the beads on. In most cases the surface antigen will be internalized after 2-3 days, and then the beads will fall off since the beads are too big to be internalized by the endocytosis pathway.

Will Dynabeads magnetic beads be internalized by the target cells?

In general, the size of the Dynabeads magnetic beads is so large that they will not be internalized. The clathrin-coated pits are typically not more than 500 nm in size, which will be too small for Dynabeads magnetic beads to be internalized by endocytosis. However, if the target cells have phagocytic activities such as monocytes/macrophages, the Dynabeads magnetic beads could be internalized by phagocytosis.

What are the recommended Dynabeads magnetic beads and cell concentrations for optimal cell isolation/depletion?

For positive cell isolation we recommend that you use 1-2 x 10E7 total PBMC per mL, and 1 x 10E7 Dynabeads magnetic beads per mL of sample. Another way of determining how many beads you need is to assume approximately 4 beads per target cell. If you want to deplete your sample of a certain target cell, we recommend using the same cell concentration as above (1-2 x 10E7 total PBMC per mL), but at least double the amount of beads to 2 x10E7 Dynabeads magnetic beads per mL of sample. Or alternatively, assume about 8-10 beads per target cell to be depleted.
Note: Most Dynabeads magnetic beads for cell separations are supplied at approximately 4 x 10E8 beads/mL, thus 1 x 10E7 beads is only 25 µL (please check the specific product manual or label for each product to confirm)

What are the pros and cons of using a positive isolation method with Dynabeads magnetic beads?

The major advantages of using positive isolation are the high purity and yield. In addition, the initial sample prep is generally not required, since the cells can be isolated directly from any sample such as whole blood, bone marrow, or tissue digests. If using the primary coated beads without bead release, the procedure is very simple and fast; for example you can get isolation of cells directly from whole blood in as little as 25 minutes, which also leads to very high cell viability. A general disadvantages for any positive isolation method is a potential activation of the cells, depending on the function of the target epitope on the target cell. Note that the Dynabeads magnetic beads are not directly causing activation; it is the antibody-cell epitope interaction that causes this. When using positive isolation with a release mechanism, some loss of yield must be expected, since not all links between the cells and beads will be broken.

What is positive isolation with Dynabeads magnetic beads?

Positive isolation is a method for separation of one specific target cell population from a sample. By using Dynabeads magnetic beads, magnetic particles coated with target-specific antibodies, the beads will bind only the cells with the matching surface antigen expressed on the target cells' surface. Two different Dynabeads magnetic beads - based positive isolation methodologies may be employed depending on the downstream assays. Since the beads themselves are quite large (from 1 µm to 4.5 µm in diameter) we don't recommended that you use bead-bound cells directly in any flow cytometer. Thus, if you have a cellular downstream assay and want to use a flow cytometer, we recommend using one of our positive isolation kits where a bead-release mechanism is available.

Can I use dPBS with calcium and magnesium to prepare isolation buffer and washing buffer?

No, the PBS must be Ca2+- and Mg2+-free as these divalent ions can lead to activation of complement and aggregation of cells. Aggregation of cells in the sample can severely reduce both yield and purity of the isolation. EDTA must also be added to the isolation buffer to minimize activation of complement and aggregation. Alternatively EDTA can be replaced by 0.6% sodium citrate.

What determines whether positive or negative isolation should be used with Dynabeads magnetic beads?

When cell purity is the most important criteria, target cell activation is not a concern, or when the downstream applications are isolation of RNA or gDNA, then positive isolation is recommended. When the most important criteria is to isolate cells with minimum disturbance, and where the downstream applications include cell culture, study cell function, morphology, and flow cytometry, then negative isolation is recommended (note that positive isolation of cells can also be used when the downstream applications include cell culture, flow cytometry, or studying cell function, but then a positive isolation with bead release from the cells are required, by using e.g., FlowComp products or the Positive Isolation Kits containing DETACHaBEAD products).

How do I thaw frozen cells?

Thaw cells in their cryovial in a 37 degrees C water bath until a small ice-clump is left. Transfer the cells gently to a fresh 10-15 mL tube immediately after the cells are thawed and add 10 mL 20% FCS/human serum in droplets to the cells while gentle pipetting. Avoid air bubbles. Work fast. Centrifuge the cells 200 X g, 8 minutes. Discard the supernatant. Resuspend in the appropriate buffer/media.

How do I freeze cells?

In general, freezing medium (10% DMSO and 90% FCS) or Gibco Recovery Cell Culture Freezing Medium (Cat. No. 12648-010) work well. Some cells will always die during the freezing process. In addition, freezing and thawing will cause some cells to lyse. The protocol to freeze mammalian cells using Gibco Recovery medium is as follows:

1. Thaw Recovery Cell Culture Freezing Medium, mix well, and keep at 2-8 degrees C until use.
2. For suspension cells proceed to step 3. For adherent cells, gently detach cells from the substrate on which they are growing using a suitable dissociation reagent such as Gibco TrypLE reagent. Resuspend cells in the complete medium required for that cell type.
3. Transfer cell suspension to a sterile 15 mL centrifuge tube.
4. Determine the viable cell density and percent viability using a Countess Automated Cell Counter (similar automated or manual methods may be used) and calculate the required volume of Recovery Cell Culture Freezing Medium to give a final cell density of 1 X 10E6 to 1 X 10E7 cells/mL.
5. Centrifuge cell suspension at 100-200 x g for 5-10 minutes. Aseptically decant supernatant without disturbing the cell pellet. Note: Centrifugation speed and duration may vary depending on cell type.
6. Resuspend the cell pellet in (2- 8 degrees C) chilled Recovery Cell Culture Freezing Medium at recommended viable cell density for specific cell type (typically 1 X 10E6 cells/mL or greater).
7. Dispense aliquots of cell suspension (mix frequently to maintain a homogeneous cell suspension) into cryovials according to the manufacturer's specifications (i.e., 1.5 mL in a 2 mL cryovial).
8. Achieve cryopreservation in an automated or manual controlled rate freezing apparatus following standard procedures (approximately 1 degree C decrease per minute).
9. Transfer frozen cells to liquid nitrogen, (vapor phase); storage at -200 degrees C to -125 degrees C is recommended.

Can bone marrow be used as starting material for cell isolation using Dynabeads magnetic beads ?

Bone marrow needs to be washed and diluted prior to addition of Dynabeads magnetic beads to make the sample less viscous. Washing and DNase treatment is recommended for preparing bone marrow cells prior to cell isolation using Dynabeads magnetic beads:

- Mix 2 mL (10E7-10E8 cells) bone marrow with 2 ml PBS w/ 0.1% BSA + 0.6% Na-citrate.
- Centrifuge at 600 g for 8 min at 18-25 degrees C.
- Discard the supernatant and resuspend to 5 mL with PBS w/ 0.1% BSA + 1mM CaCl2 + 0.5 mM MgCl2.
- Add 600 Kunitz units DNase I (120 Kunitz units DNase I per milliliter).
- Incubate cells for 30 minutes at 18-25 degrees C with both gentle tilting and rotation.
- Centrifuge cell suspension for 8 minutes, 600 x g, at 18-25 degrees C.
- Discard supernatant and resuspend cell pellet in 5 mL PBS w/ 0.1% BSA.
- Centrifuge cell suspension for 8 minutes, 600 x g, at 18-25 degrees C.
- Discard supernatant and resuspend at 1 x 10E8 cells per milliliter in RPMI 1640 / 1% FCS

How do I carry out tissue digests to obtain a single-cell suspension before using Dynabeads magnetic beads for cell isolation?

Follow standard tissue preparations using enzymes and mechanical disruption to get a single-cell suspension. Eliminate large aggregates by sieving the digested cell suspension through a cell strainer or filter through a 30 µm filter. Disruption of tissue normally results in some cell death and release of DNA. Free DNA will impair cell capture, recovery, and purity. DNase I treatment is performed by incubating the cell suspension in PBS with 0.1% BSA + 1 mM CaCl2 + 0.5 mM MgCl2 and 120 Kunitz units DNase I per ml at 18-25 degrees C for 30 min. (For CELLection products, wash cells to remove DNase before adding the beads.)

How do I prepare mononuclear cells (MNC), and what kind of cells are present in MNC and in what proportion?

Mononuclear cells (MNC), also known as peripheral blood mononuclear cells (PBMC), are prepared from whole blood, buffy coat, bone marrow, or umbilical cord blood by density gradient separation. The following protocol can be used for standard MNC preparation for positive isolation or depletion protocols:

1. Collect blood sample with anticoagulant present (EDTA, ACD, heparin). Dilute peripheral blood 1 + 1, buffy coat 1 + 2, bone marrow 1 + 1 and umbilical cord blood 1 + 3 in PBS w/ 0.1% BSA + 0.6% Na-citrate or 2 mM EDTA.
2. Layer up to 35 mL of the diluted sample over 15 mL gradient medium (such as Ficoll or Lymphoprep solution) in a 50 mL tube.
3. Centrifuge for 400 x g for 30-40 minutes at 18-20 degrees C. If blood has been stored for more than 2 hours, increase centrifugation time by 10 min.
4. Collect MNC from the interface and transfer cells to a 50 mL tube.
5. Wash MNC three times with PBS w/0.1% BSA by centrifugation at 300 x g for 8 min at 2-8 degrees C.
6. Resuspend the cells to 1 x 10E7 cells per milliliter in PBS with 0.1% BSA and cool to 2-8 degrees C.
Note: MNC contain T cells (50%), B cells (5-10%), NK cells (5-10%), and monocytes (30%) without granulocytes and very few platelets.

For use with Untouched/negative isolation kits, the following protocol is recommended to obtain MNC prep with low platelet numbers and the highest possible purity:

Whole blood/buffy coat and bone marrow can be used as a starting material.

1. Dilute 10-18 mL blood/buffy coat with PBS w/ 0.1% BSA + 0.6% Na-citrate or 2 mM EDTA to a total volume of 35 ml at 18-25 degrees C.
2. Add the diluted blood/buffy coat on top of 15 mL of gradient medium (such as Lymphoprep or Ficoll solution).
3 .Centrifuge at 160 x g for 20 min at 20 degrees C. Allow to decelerate without braking.
4. Remove 20 mL of supernatant to eliminate platelets.
5. Centrifuge at 350 x g for 20 min at 20 degrees C. Allow to decelerate without braking.
6 .Recover MNC from the plasma/Lymphoprep solution interface and transfer the cells to a 50 mL tube.
7. Wash MNC once with PBS w/ 0.1% BSA by centrifugation at 400 x g for 8 min at 2-8 degrees C.
8. Wash MNC twice with PBS w/ 0.1% BSA by centrifugation at 225 x g for 8 min at 2-8 degrees C and resuspend the MNC at 1 x 10E8 MNC per milliliter in PBS w/ 0.1% BSA.

What is a buffy coat?

Buffy coat, also known as leukocyte concentrate, is the middle fraction of an anti-coagulated blood sample that sits under the plasma and on the top of red blood cells after centrifugation of the sample without using a density gradient reagent such as Ficoll solution. Buffy coat contains both leukocytes and platelets and can be used as a source of this cellular material.

What is the concentration of major cell populations in normal adult human blood?

Typically, one milliliter of adult human blood contains:
~5 x 10E9 red blood cells
~7 x 10E6 leukocytes
~3 x 10E8 platelets

In the 7 x 10E6 leukocyte fraction, there are:
4 x 10E5 monocytes
1 x 10E5 NK cells

Lymphocytes:
2 x 10E5 B cells
1 x 10E6 T cells (approx. 70% are CD4+ T cells and 30% are CD8+ T cells)

Granulocytes:
5 x 10E6 neutrophils
2 x 10E5 eosinophils
4 x 10E4 basophils

What Dynabeads magnetic bead size should I select for my application?

This will depend on your application. As a guideline, the 4.5 micron beads are best used for cell isolation and activation/expansion. These larger beads have a higher magnetic mobility, they are roughly the same size as mammalian cells, and are less likely to be taken up by the cells. The smaller 1 micron beads and 2.8 micron beads are often used when isolating nucleic acids or proteins, or for immunoprecipitation. In negative cell isolation kits, one micron beads are often used because of their higher binding capacity per milliliter of beads and faster binding kinetics. With negative selection, cells taking up any beads will not be a problem as you want to look at the remaining cell population anyway. The 2.8 micron Dynabeads magnetic beads, coated with secondary antibodies, protein A or protein G, or streptavidin are also used for positive cell isolation with primary antibodies of your own choice, targeting specific cell-surface antigens.

How large are the Dynabeads magnetic beads?

Three different sizes of Dynabeads magnetic beads are available: One micron beads (look for MyOne magnetic beads in the product name), 2.8 micron beads, and 4.5 micron beads. In general, the binding capacity per milliliter of beads and binding kinetics increases as the bead size reduces.

What are the characteristics and advantages of Dynabeads magnetic beads?

Dynabeads magnetic beads are super-paramagnetic, meaning they only display magnetic characteristics when a magnet is present. As soon as the magnet is removed, the beads handle like a liquid and are easily dispersed in the sample tube. For cell isolation purposes, this has clear advantages as it allows for gentle handing and reduced stress to the cells. Secondly, the beads all have the same size and shape, with rapid liquid-phase reaction kinetics. The smooth surface of the beads results in less non-specific binding. These properties tend to reduce variability and allow you to get more reliable and reproducible results for your purifications and your analyses whether you are looking at cells or any other target molecule (RNA/DNA/proteins/protein complexes/organelles/exosomes etc.)

Can you please offer some tips to improve the releasing efficiency with Dynabeads FlowComp cell isolation kits?

Here are some suggestions:

- Since the DSB-X biotin -streptavidin bond becomes stronger over time, do not increase the time of the release step longer than stated in the protocol. In some instances a shorter incubation time can lead to higher yield.
- After incubation with the releasing agent, we recommend that you resuspend the sample well by pipetting >10 times using a 1 mL pipette before adding the sample to the magnet.

What can I do to improve cell yield when isolating cells with any of the Dynabeads positive isolation kits?

Here are some suggestions:

- Sample preparation is very important. All isolations must be performed on single-cell suspensions. For isolation of certain cell types (e.g., monocytes) directly from whole blood, we recommend that you wash the blood before isolation to remove interfering factors in the sample.
- An appropriate mixer must be used for all incubations where ‘mixing' is specified. Any mixer providing either tilting and rotation or end-over-end mixing can be used (e.g., HulaMixer Sample Mixer (Cat. No. 15920D)).
- If using FlowComp kits where the antibodies are added to the cells before adding the beads (indirect method), excess antibodies should be removed by washing prior to adding the beads.
- After incubation with the releasing agent, we recommend that you re-suspend the sample well by pipetting >10 times using a 1 mL pipette before adding the sample to the magnet.
- The recommended isolation buffer must be used for cell isolations. The PBS must be Ca2+ and Mg2+ free as these divalent ions can lead to activation of complement and aggregation of cells.
- Aggregation of cells in the sample can severely reduce both yield and purity of the isolation.


For Research Use Only. Not for use in diagnostic procedures.