Learn More
Invitrogen™ ChargeSwitch™-Pro Plasmid Miniprep Kit
The ChargeSwitch™-Pro Plasmid Miniprep Kit generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications.
$202.00
Specifications
| Column Type | Spin Column |
|---|---|
| Content And Storage | Store at room temperature. |
| Format | Kit |
| Isolation Technology | ChargeSwitch Chemistry |
| Sample Type | Bacterial Culture |
Includes: Miniprep Columns with Collection Tubes, Recovery Tubes, Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffers 1 and 2, and Elution Buffer
Description
The ChargeSwitch™-Pro Plasmid Miniprep Kit generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications.
• 100% ethanol-free, guanidinium-free, organics-free, aqueous protocol for a safer, more streamlined method
• Cleaner purified product for improved performance in downstream applications
• Ready-to-use, complete kit for a fast, hassle-free start
• Ideal for purifying plasmid DNA from 1-5 mL overnight bacterial cultures, yielding up to 20 μg of plasmid DNA
Order Info
Shipping Condition: Room Temperature
Specifications
| Spin Column | |
| Kit | |
| Bacterial Culture | |
| Not High-throughput Compatible (Manual) | |
| ChargeSwitch | |
| Room Temperature |
| Store at room temperature. | |
| ChargeSwitch Chemistry | |
| Plasmid DNA | |
| < 100 μg (Small-Scale) Plasmid DNA | |
| Plasmid MiniPrep Kit | |
| Plasmid DNA |
For Research Use Only. Not for use in diagnostic procedures.