missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BioPrime™ DNA Labeling System

Catalog No. 18094011
Encompass
Change view
Click to view available options
Quantity:
30 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18094011 30 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18094011 Supplier Invitrogen™ Supplier No. 18094011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

700μL 2.5X Random Primers Solution; 175μL 10X dNTP Mixture; 100μg Control DNA; 35μL Klenow Fragment (40U/μL); 500μL Stop Buffer; 1.25mL Distilled Water

Specifically designed for use in the preparation of biotinylated probes

Random primers (octamers) are annealed to the denatured DNA template and extended by Klenow fragment in the presence of biotin-14-dCTP to produce sensitive biotinylated-DNA probes for use in the nonradioactive detection of DNA and RNA. With the BioPrime™ DNA Labeling System, considerable net DNA synthesis occurs resulting in a 10–40 fold amplification of the probe. Probes made with this product have been used for hybridization to Southern and northern blots, plaque and colony lifts, and for in situ hybridization to chromosome spreads.

  • Particularly well suited for situations in which the DNA is limited
  • Requires less template DNA than nick translation
  • Amplifies template to provide an increased amount of biotinylated probe
  • Labels 50 to 500ng of template DNA in one reaction

Array CGH, DNA and RNA Purification and Analysis, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Southern Blotting

Order Info

Shipping Condition: Approved for shipment on Wet or Dry Ice

Specifications

Content And Storage • 700 μl 2.5X Random Primers Solution
• 175 μl 10X dNTP Mixture
• 100 μg Control DNA
• 35 μl Klenow Fragment (40 U/μl)
• 500 μl Stop Buffer
• 1.25 ml Distilled Water

Store at -20°C in a non-frost-free freezer.
Detection Method Biotin-based
Label or Dye Biotin
Final Product Type Probes (Labeled DNA)
Labeling Target DNA (General), Genomic DNA
Includes Label or Dye Yes
Labeling Method Direct Labeling
Product Line BioPrime
Product Type DNA Labeling System
Quantity 30 Reactions
Shipping Condition Approved for shipment on Wet or Dry Ice
Show More Show Less
Can exo-Klenow be purchased separately from your array labeling kits?

Yes, the catalog number for the exo-Klenow is AM2008. The enzyme concentration is 5 U/µL.

For Research Use Only. Not for use in diagnostic procedures.