missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 2-YT Broth

Catalog No. 22712020
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22712020 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22712020 Supplier Invitrogen™ Supplier No. 22712020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2-YT Broth is a nutrient-rich microbial growth medium used for cultivation of E. coli and growth of hosts for replication of M13 vectors.

2-YT Broth is a nutrient-rich microbial growth medium used for cultivation of E. coli and growth of hosts for replication of M13 vectors. The 2-YT formulation is similar to Lennox LB medium, but contains double the concentration of yeast extract and a 60% higher concentration of peptone. 2-YT medium allows cells to reach higher density and increase recombinant protein production compared to LB media formulations.

2-YT Broth formulation
Formulation per liter: 16 g SELECT Peptone 140 (pancreatic digest of casein), 10 g SELECT Yeast Extract, and 5 g NaCl.
Note: To prepare 1 L of 2-YT medium, weigh 31 g of powder and add distilled water to 1 L. Adjust the pH value to 7.0 with 5 N NaOH and sterilize by autoclaving for 20 minutes at 121°C.

Primary applications
• Growth of hosts for replication of M13 vectors
• Recombinant protein expression
• Achieve higher plasmid DNA yield

Find the media format that fits you needs
We offer a variety of ready-to-use media formulations including LB Broth (Cat. No. 10855001), Terrific Broth (Cat. No. A1374301) and S.O.C Recovery Medium (Cat. No. 15544034).

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage • 2-YT Broth Powder (500 g)

Store at room temperature.

Format Bottle
Media Type 2-YT Broth (Yeast Extract Tryptone)
Preparation Method Autoclave
Product Type Dehydrated Culture Media
Target Organism Class Coliforms
Quantity 500 g
Shipping Condition Room Temperature
Species Escherichia coli

For Research Use Only. Not for use in diagnostic procedures.