missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ SuperFi™ PCR Master Mix

Catalog No. 12358050
Encompass
Change view
Click to view available options
No. of Reactions:
100 Reactions
5 x 500 Reactions
500 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
12358050 500 Reactions
12358010 100 Reactions
12358250 5 x 500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12358050 Supplier Invitrogen™ Supplier No. 12358050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen Platinum SuperFi PCR Master Mix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification.

Invitrogen Platinum SuperFi PCR Master Mix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification. Only template, primers, and water need to be added, thus reducing the number of pipetting steps during PCR reaction setup.

Platinum SuperFi PCR Master Mix retains all the features of Platinum SuperFi DNA Polymerase, a proofreading DNA polymerase that combines superior fidelity with trusted Platinum hot-start technology. Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis, and other applications benefiting from supreme sequence accuracy.

Platinum SuperFi DNA Polymerase is engineered with a DNA-binding domain resulting in high processivity and increased resistance to PCR inhibitors. This feature also enables fast-cycling protocols and amplification of long targets. The Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation.

Features

  • Exceptional >300X Taq fidelity
  • High specificity and increased yields with Platinum hot-start technology
  • Robust amplification of difficult targets of high GC content
  • Room temperature reaction setup and 24-hour bench top stability of pre-assembled reactions
  • Amplification of samples with suboptimal purity

Applications

  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates
  • Template generation for sequencing
  • High-throughput PCR
  • Long PCR

Notes

  • Annealing rules for Platinum SuperFi DNA Polymerase are different from many common DNA polymerases.
  • Platinum SuperFi PCR Master Mix is supplied with separate SuperFi GC Enhancer for difficult and GC-rich templates (˃65% GC).

Specifications

Content And Storage • 2X Platinum SuperFi PCR Master Mix (10 x 1.25 mL)
• SuperFi GC Enhancer (4 x 1.25 mL)
• Nuclease-free water (10 x 1.25 mL)

Store at –5°C to –30°C.
GC-Rich PCR Performance High
Polymerase Platinum SuperFi DNA Polymerase
Reaction Speed Fast
Quantity 500 reactions
Shipping Condition Wet or Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) 300X
Hot Start Built-In Hot Start
No. of Reactions 500 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Size (Final Product) 20 kb or less
Show More Show Less
Which nucleotide analogues can be used with Platinum SuperFi DNA Polymerase?

Platinum SuperFi DNA Polymerase cannot read dUTP-derivatives or dITP in DNA templates, so the use of these analogues is not recommended.Platinum SuperFi DNA Polymerase can incorporate 7-deaza-dGTP and radiolabeled dNTPs. 

I am having difficulties amplifying long targets using Platinum SuperFi DNA Polymerase. What are your recommendations?

Good lab practices are important for long fragment amplification. These include using high-quality templates (pure, fresh and intact) and fresh primer solutions. Optimization steps to consider include decreasing denaturation and extension temperatures, lengthening extension times as recommended in the manual, and increasing template amounts.

Can Platinum SuperFi DNA Polymerase amplify AT-rich targets?

Yes. To improve amplification of AT-rich targets, we recommend reducing the extension temperature to 68 degrees C or add 5-15 mM Tetramethylammonium Chloride (TMAC).

Can Platinum SuperFi DNA Polymerase amplify GC-rich targets?

All Platinum SuperFi product formats are supplied with 5X SuperFi GC Enhancer which is optimized to improve amplification of GC-rich targets (recommended for use with targets containing >65% GC content)

Does SuperFi Green Buffer influence performance of Platinum SuperFi DNA Polymerase?

No. The colored buffer does not interfere with PCR performance and does not change enzyme features.

Can Platinum SuperFi DNA Polymerase be used for amplification of bisulfite-converted DNA?

No. Platinum SuperFi DNA Polymerase cannot read uracil in the template strand, therefore, it is not recommended for use with bisulfite-converted DNA.

Does Platinum SuperFi DNA Polymerase add the non-template dependent 3'-A overhang?

No. Platinum SuperFi DNA Polymerase generates blunt ended products.

What is the amplicon size limit using Platinum SuperFi DNA Polymerase?

Platinum SuperFi DNA Polymerase accurately amplifies long fragments (up to 20 kb) with high yields and specificity. Amplification of even larger fragment sizes up to 40 kb has been demonstrated, but may require additional optimization of reaction conditions and primer design.

Can protocols optimized for Taq DNA Polymerase, Platinum Taq DNA Polymerase High Fidelity, AccuPrime Taq DNA Polymerase High Fidelity, Platinum Pfx DNA Polymerase, or AccuPrime Pfx DNA Polymerase be directly applied to Platinum SuperFi DNA Polymerase?

Platinum SuperFi DNA Polymerase significantly differs from many other DNA polymerases, therefore annealing rules should be adjusted by using our Tm calculator (www.thermofisher.com/tmcalculator). Due to high processivity, Platinum SuperFi DNA Polymerase also has shorter cycling times than many other DNA polymerases.


For Research Use Only. Not for use in diagnostic procedures.