missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inulin, Spectrum™ Chemical
Spécifications
Spécifications
| CAS Max % | 1 |
| Loss on Drying | 0.1 |
| Packaging | Amber Glass Bottle |
| Quantity | 5 g |
| Grade | Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.