missing translation for 'onlineSavingsMsg'
Learn More

ZOLL™ Intellisense™ Battery For Powerheart™ G5 AED

Catalog No. 17478089
Change view
Click to view available options
Product Type:
Battery
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17478089 Battery
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17478089 Supplier ZOLL™ Supplier No. XBTAED001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Medical grade battery for Powerheart™ G5 AED

  • Communicates performance condition, age, storage temp and vital info
  • Four year replacement program

Compatible With:

  • Powerheart™ G5

Notes:

  • Use of after-market batter will void AED warranty

Specifications

Product Type Battery
Color Orange
For Use With (Equipment) Powerheart™ G5 AED