Learn More
Perkin Elmer Canada Integrated Platform THGA Graphite Tubes with End-Cap, Pkg. 10
Numéro de catalogue. 502113427
Encompass
Made from PerkinElmer's own blend of high-quality base graphite material. This provides the user with the required performance to yield reproducible results from day to day with respect to sensitivity (characteristic mass), accuracy and precision.
Numéro de catalogue. 502113427 Fournisseur Perkin Elmer Canada Code fournisseur N9304643
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Made from PerkinElmer's own blend of high-quality base graphite material. This provides the user with the required performance to yield reproducible results from day to day with respect to sensitivity (characteristic mass), accuracy and precision.