Learn More
Integra™ Miltex™ Microsurgical Kit
Kit with a selection of commonly used surgical tools
Spécifications
| For Use With (Equipment) | Lab animal |
|---|
Exige:
Kit includes one each:
- DERF needle holder with grated jaws, 4000 teeth per square in
- 4 in. long (10.2cm) Iris scissors
- 4 in. long (10.2cm) Swiss jeweler style forceps, nonmagnetic stainless steel, style 5, micro-fine
- 3.5 in. long (8.9cm) McPherson micro iris suturing forceps, straight, smooth 0.3 x 5mm platform, no teeth
- 3.25 in. long (8.3cm) McPherson-Vannas micro Iris scissors, straight with sharp points
| Numéro de catalogue | Numéro du manufacturier. | Product Type | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Product Type | Prix | Quantity & Availability | |||||
|
12460112
|
Integra™ Miltex™
6890 |
Microsurgical Lab Animal Kit w/ Storage Cassette and Mat |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Available with or without stainless-steel storage cassette and mat.
†Supplied only with Cat. No. 12-460-112.
Spécifications
| Lab animal |