missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Inositol hexaphosphoric acid, 50 wt% aqueous solution

Catalog No. AC235372500
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC235372500 250 mL
AC235370010 1 L
AC235370025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC235372500 Supplier Thermo Scientific Chemicals Supplier No. 235372500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 83-86-3 | C6H18O24P6 | 660.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Inositol hexaphosphoric acid
Name Note 40-50 wt. % Aqueous Solution
CAS 83-86-3, 7732-18-5
Boiling Point 105.0°C
Molecular Formula C6H18O24P6
MDL Number MFCD00082309
Density 1.3700g/mL
Synonym phytic acid, phytate, fytic acid, inositol hexaphosphate, alkalovert, myo-inositol hexaphosphate, alkovert, phytine, acide fytique, acidum fyticum
Solubility Information Solubility in water: miscible.
InChI Key IMQLKJBTEOYOSI-GPIVLXJGSA-N
SMILES OP(O)(=O)O[C@H]1[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@@H]1OP(O)(O)=O
IUPAC Name (2,3,4,5,6-pentaphosphonooxycyclohexyl) dihydrogen phosphate
Molecular Weight (g/mol) 660.03
PubChem CID 890
Formula Weight 660.03
Packaging Glass bottle
Refractive Index 1.391
Specific Gravity 1.37
Quantity 250 mL
Show More Show Less

RUO – Research Use Only