missing translation for 'onlineSavingsMsg'
Learn More

Inorganic Carbon Standard, Reagents

Inorganic Carbon Standard, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc CC2058001A

 View more versions of this product

Catalog No. 01-335-583


Only null left

Description

Description

Specifications

Specifications

Inorganic Carbon Standard
7732-18-5 , 497-19-8
Liquid
100°C
Completely miscible
1 L
Poly Bottle
1.0045
Colorless
Water Analyzer
Water Analysis
Reagent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions