missing translation for 'onlineSavingsMsg'
Learn More

Inorganic Carbon Standard, Reagents

Catalog No. p-200022064
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335584 125 mL
01335583 1 L
01335582 4 L
01335581 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01335584 Supplier Labchem Inc Supplier No. CC205800125A
Only null left

Inorganic Carbon Standard, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Inorganic Carbon Standard
Packaging Poly Bottle
CAS 7732-18-5, 497-19-8
Specific Gravity 1.0045
Physical Form Liquid
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Water Analysis
Quantity 125 mL
Grade Reagent
Show More Show Less