missing translation for 'onlineSavingsMsg'
Learn More

Inorganic Carbon Standard, 500 ppm C, Ricca Chemical

Catalog No. R18455001C
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R18455001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R18455001C Supplier Ricca Chemical Company Supplier No. R18455001C

Chemical Identifiers

CAS 144-55-8 , 497-19-8 , 7732-18-5
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Packaging Amber Glass Bottle
Odor Odorless
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Miscible with water
Solution Type Calibration Solution
Type TOC Standard
Quantity 1 L
Grade Laboratory
Chemical Name or Material Inorganic Carbon Standard, 500 ppm C
Show More Show Less
ShelfLife 12 months