missing translation for 'onlineSavingsMsg'
Learn More

Inorganic Carbon Standard, 25 ppm C, Ricca Chemical

Catalog No. R1845025250
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R1845025250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R1845025250 Supplier Ricca Chemical Company Supplier No. R1845025250C

Chemical Identifiers

CAS 144-55-8 , 497-19-8 , 7732-18-5
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Packaging Amber Glass Bottle
Odor Odorless
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Miscible with water
Solution Type Calibration Solution
Type TOC Standard
Quantity 250 mL
Grade Laboratory
Chemical Name or Material Inorganic Carbon Standard, 25 ppm C
Show More Show Less
ShelfLife 12 months