missing translation for 'onlineSavingsMsg'
Learn More

Inorganic Carbon Standard, 100 ppm C, Ricca Chemical

Catalog No. R1845100500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R1845100500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R1845100500 Supplier Ricca Chemical Company Supplier No. R1845100500C1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 144-55-8 , 497-19-8 , 7732-18-5
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Packaging Safety-coated Glass Bottle
Odor Odorless
Assay Percent Range 100 ± 1ppm C
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Infinitely soluble in water
Solution Type Calibration Standard
Type Analytical Standard
Solvent or Matrix Water
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Inorganic Carbon Standard, 1mL = 1mg C, 100ppm C
Show More Show Less