Learn More
Alkali Scientific Inoculating Loops, 10ul, Flexible PP (50 Sterile Peel Bags of 20 Loops), 1000/cs
Numéro de catalogue. 502335761
Encompass
These sterile, 10ul, disposable plastic loops are designed for use in quantitative procedures such as sampling, urine counts and serial dilutions, as well as for bacterial inoculation. Because they are pre-sterilized and designed for single use, there is no risk of cross contamination and no need for flaming. These inoculating loops are flexible enough for streaking, yet rigid enough for subsurface inoculation. The unique hexagonal shape of the shaft makes it easier to grip, orientate and guide through a smooth streaking process. The loop surfaces are extremely smooth with no rough edges and no flashing or burrs on the loop head. This results in the elimination of cutting or gouging of the agar surface. sterile, disposable and verifiably accurate, these inoculation loops provide an efficient, cost-effective and time-saving alternative to traditional metallic loops.
Numéro de catalogue. 502335761 Fournisseur Alkali Scientific Code fournisseur M2025
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
These sterile, 10ul, disposable plastic loops are designed for use in quantitative procedures such as sampling, urine counts and serial dilutions, as well as for bacterial inoculation. Because they are pre-sterilized and designed for single use, there is no risk of cross contamination and no need for flaming. These inoculating loops are flexible enough for streaking, yet rigid enough for subsurface inoculation. The unique hexagonal shape of the shaft makes it easier to grip, orientate and guide through a smooth streaking process. The loop surfaces are extremely smooth with no rough edges and no flashing or burrs on the loop head. This results in the elimination of cutting or gouging of the agar surface. sterile, disposable and verifiably accurate, these inoculation loops provide an efficient, cost-effective and time-saving alternative to traditional metallic loops.