missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Inoculating Nichrome Loops 8 in. Handle

Fisherbrand™ Innoculating loops are available in a variety of gauges, loop hole sizes, and wire types to accommodate an array of laboratory requirements.

Supplier:  Fisherbrand™ 10401129

 View more versions of this product

Catalog No. 13820028



Only null left

Description

Description

Fisherbrand™ Nichrome/Aluminum Innoculating Loops consist of a nichrome wire loop mounted to an aluminum handle. The aluminum handle is available in either an insulated or non-insulated format.

  • Ideal for selecting, inoculating, or transferring microbiological samples
  • Cools rapidly after sterilization
  • Loops Slide along agar surfaces without cutting or tearing material
  • Nichrome wire available in single or twisted configuration
  • Passivated
  • 203 mm (8 in.) handles, available with insulation
  • Compatible with Gamma/ETO/Autoclavable sterilization methods

Certifications

ISO 9001-02, GMP, cGMP

Specifications

Chemical Identifiers

11 in.
Nichrome
Inoculating Loop
Available as Each or Pk. of 12
Ideal for selecting, inoculating, or transferring microbiological samples
280 mm
Silver
Petri-Dish etc
Autoclavable

Specifications

11 in.
Nichrome
Silver
Petri-Dish etc
Nichrome wire with aluminum handle
Autoclavable
280 mm
3 mm
Inoculating Loop
25
Available as Each or Pk. of 12
Ideal for selecting, inoculating, or transferring microbiological samples
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions