missing translation for 'onlineSavingsMsg'
Learn More

Surgical Design™ Inoculating Loops with Aluminum Handles

Surgical Design™ Inoculating Loops with Aluminum Handles are used to cultivate microbes on plates by transferring inoculum for streaking.

Supplier:  Surgical Design™ D28645

 Afficher plus de versions de ce produit

Numéro de catalogue. 22079728



Il en reste null

Description

Description

  • Ideal for selecting, inoculating, or transferring microbiological samples
  • Cools rapidly after sterilization
  • Smooth loops for damage prevention to sample
  • Reusable loops feature plain aluminum handle
  • Available in single or braided wire
  • Loop sizes: 2mm, 3mm, 4mm, 5mm
Spécifications

Identifiants chimiques

Nichrome
144
Autoclavable
Inoculation

Spécifications

Nichrome
Single Wire
Autoclavable
3 mm
144
Inoculation
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions