Research Products Corporation Innoculating Loop, 1 ul, Blue, Sterile, 25 per Package, 500 per Case
Numéro de catalogue. 50476473
Inoculating loops and needles are smooth and flexible for uniform streaking without damaging the gel surface. Made of durable polystyrene for easy disposal after each use. Supplied in sealed packs with certification. Packaged 25 per pack, 500 per case.
Numéro de catalogue.
50476473
Fournisseur
Research Products CorporationCode fournisseur
RPI 161210
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Inoculating loops and needles are smooth and flexible for uniform streaking without damaging the gel surface. Made of durable polystyrene for easy disposal after each use. Supplied in sealed packs with certification. Packaged 25 per pack, 500 per case.