Learn More
Mettler Toledo™ InLab™ OptiOx Pro-DES™ DO EasyPlace Adapter
Description
- Use the adapter to install the InLab OptiOx Pro-DES sensor into the EasyPlace electrode arm.
- For use with METTLER TOLEDO NineFocus and InLab OptiOx Pro-DES digital sensor
Specifications
Specifications
| Type | Probe Holder |