Learn More
Mettler Toledo™ InLab™ OptiOx Pro-DES™ DO BOD Adapter
Description
- Use the BOD adapter to install the InLab OptiOx Pro-DES sensor on all common BOD applications.
Spécifications
Spécifications
| Catégorie | BOD Adapter |