Learn More
Mettler Toledo™ InLab™ DES™ Cable
Numéro de catalogue. 01915888
Modifier l'affichage
Click to view available options
Length (Metric) Cable:
1.2 m
3 m
5 m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Length (Metric) Cable
01915888 1.2 m
01915875 3 m
01915876 5 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 01915888 Fournisseur Mettler Toledo™ Code fournisseur 31052331
Digital cable designed for secure connection with DES sensors, ensuring stable signal transmission and reliable measurement results.
  • One cable for all digital parameters
  • DES™ - Diagnosable Efficient Sustainable:
    • Increased efficiency with sensor diagnostics
    • Faster problem solving, less downtime, maximized sensor lifespan, and support of preventive maintenance.
  • Stay ahead: The sensor's diagnostic capability monitors sensor condition, supports preventive maintenance, maximizes the sensor's lifespan, and prevents premature discard
  • Sustainable investment: DES™ digital sensors are a smart choice. Retaining the measurement electronics and only replacing the sensor shaft at the end of its life, or the detachable cable that can be used for all sensors, helps reduce material waste and thus, the ecological footprint
  • Boost productivity and efficiency: With diagnosed errors, the user guidance with troubleshooting recommendations enables faster problem-solving, reducing downtime. It also minimizes handling errors, thereby cutting down on the need for specialized training
Spécifications
Type Cable
Length (Metric) Cable 1.2 m