missing translation for 'onlineSavingsMsg'
Learn More

Iniparib, Thermo Scientific™

Catalog No. p-7238598
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469161000 100 mg
AC469165000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC469161000 Supplier Thermo Scientific Chemicals Supplier No. 469161000
Only null left

Chemical Identifiers

CAS 160003-66-7
Molecular Formula C7 H5 I N2 O3

Specifications

Quantity 100 mg
Formula Weight 298.30
MDL Number MFCD11110639
Physical Form Powder
Color White to Beige
Purity ≥97.5%
InChI Key MDOJTZQKHMAPBK-UHFFFAOYSA-N
SMILES NC(=O)C1=CC(=C(I)C=C1)[N+]([O-])=O
IUPAC Name 4-iodo-3-nitrobenzamide
Molecular Weight (g/mol) 292.03
Chemical Name or Material Iniparib
Show More Show Less
Hazard Category Exclamation mark
Recommended Storage Normal conditions