accessibility menu, dialog, popup

UserName

CAS RN 160003-66-7

IUPAC Name: 4-iodo-3-nitrobenzamide
Synonyms: iniparib
Molecular Weight (g/mol): 292.03
Molecular Formula: C7H5IN2O3
InChi Key: MDOJTZQKHMAPBK-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(=C(I)C=C1)[N+]([O-])=O
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)