Learn More
Thermo Scientific Chemicals Inhibitor Removal Resin
CAS: 9003-70-7 | C24H27NO5 | 409.48 g/mol
Supplier: Thermo Scientific Chemicals 04248936
Description
Inhibitor removal resin is used for the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol. It is also take part in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Inhibitor Removal Resin | |
| Odorless | |
| 9003-70-7 | |
| For the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol | |
| CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C=O | |
| 409.48 |
| 9003-70-7 | |
| C24H27NO5 | |
| Crystalline Solid | |
| BCIPGSZQUDLGSY-UHFFFAOYNA-N | |
| tert-butyl 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoate | |
| 500 g |
Chemical Identifiers
| 9003-70-7 |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only