missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Inhibitor Removal Resin

Catalog No. p-7070056
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
AA4248914 25 g
AA4248922 100 g
AA4248936 500 g
3 options

Catalog No. AA4248914

Supplier: Thermo Scientific Chemicals 04248914

Only null left

CAS: 9003-70-7 | C24H27NO5 | 409.48 g/mol

Inhibitor removal resin is used for the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol. It is also take part in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9003-70-7

Specifications

Product Type Inhibitor Removal Resin
Odor Odorless
Molecular Formula C24H27NO5
MDL Number 9003-70-7
Form Crystalline Solid
For Use With (Application) For the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol
InChI Key BCIPGSZQUDLGSY-UHFFFAOYNA-N
SMILES CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C=O
IUPAC Name tert-butyl 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoate
Molecular Weight (g/mol) 409.48
Quantity 25 g
Show More Show Less
Recommended Storage Ambient temperatures

RUO – Research Use Only