missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Inhibitor Removal Resin

CAS: 9003-70-7 | C24H27NO5 | 409.48 g/mol

Supplier:  Thermo Scientific Chemicals 04248922

 View more versions of this product

Catalog No. AA4248922


Only null left
Explore available promotions

Description

Description

Inhibitor removal resin is used for the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol. It is also take part in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Inhibitor Removal Resin
Odorless
9003-70-7
For the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol
CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C=O
409.48
9003-70-7
C24H27NO5
Crystalline Solid
BCIPGSZQUDLGSY-UHFFFAOYNA-N
tert-butyl 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoate
100 g

Chemical Identifiers

9003-70-7
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only